Skip to Content
Merck
All Photos(1)

Key Documents

149225

Sigma-Aldrich

Dipropyl disulfide

98%

Synonym(s):

Propyl disulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2SSCH2CH2CH3
CAS Number:
Molecular Weight:
150.31
Beilstein:
969200
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.497 (lit.)

bp

195-196 °C (lit.)

mp

−86 °C (lit.)

density

0.96 g/mL at 25 °C (lit.)

SMILES string

CCCSSCCC

InChI

1S/C6H14S2/c1-3-5-7-8-6-4-2/h3-6H2,1-2H3

InChI key

ALVPFGSHPUPROW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dipropyl disulfide (DPDS) was an efficient scavenger of reactive oxygen species (ROS) at a lower concentration in both HL-60 and HepG2 cells. It prevents the oxidative DNA damage caused by N-nitrosopiperidine (NPIP) and N-nitrosodibutylamine (NDBA).

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

147.2 °F - closed cup

Flash Point(C)

64 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Han-Seung Shin et al.
Journal of agricultural and food chemistry, 50(26), 7684-7690 (2002-12-12)
Organosulfur compounds and sodium bisulfite significantly inhibited (P < 0.05) heterocyclic aromatic amine (HAA) formation in model systems containing phenylalanine, creatinine, and glucose. There was, however, no inhibition by the same compounds in a model system containing only phenylalanine and
C Teyssier et al.
Drug metabolism and disposition: the biological fate of chemicals, 28(6), 648-654 (2000-05-23)
The metabolism of dipropyl disulfide (DPDS), an Allium sulfur compound, was investigated in rat liver cell subfractions and in an isolated perfused rat liver. DPDS is oxidized to dipropyl thiosulfinate (DPDSO) by rat microsomes. The contribution of cytochrome P450 enzymes
D Guyonnet et al.
Toxicology and applied pharmacology, 154(1), 50-58 (1999-01-12)
The naturally occurring organosulfur compounds (OSCs) diallyl sulfide (DAS), diallyl disulfide (DADS), dipropyl sulfide (DPS), and dipropyl disulfide (DPDS) were studied with respect to their effects on hepatic, intestinal, renal, and pulmonary phase II drug metabolizing enzymes, i.e., glutathione S-transferase
S K Srivastava et al.
Cancer letters, 118(1), 61-67 (1997-10-06)
The mechanism of differential efficacies of diallyl sulfide (DAS), diallyl disulfide (DADS), diallyl trisulfide (DATS), dipropyl sulfide (DPS) and dipropyl disulfide (DPDS) in preventing benzo(a)pyrene (BP)-induced cancer in mice has been investigated by determining their effects on the enzymes of
D Guyonnet et al.
Mutation research, 495(1-2), 135-145 (2001-07-13)
In a previous study, we showed that naturally occurring organosulfur compounds (OSCs) from garlic and onion modulated the activation of carcinogen via the alteration of cytochromes P450. The present study was undertaken to determine the incidence of the in vivo

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service