Skip to Content
Merck
All Photos(1)

Key Documents

47517

Sigma-Aldrich

Fmoc-Hyp(tBu)-OH

≥98.0% (HPLC)

Synonym(s):

Fmoc-O-tert-butyl-L-hydroxyproline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H27NO5
CAS Number:
Molecular Weight:
409.47
Beilstein:
6161467
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

Assay

≥98.0% (HPLC)

optical activity

[α]20/D −25±2.5°, c = 1% in methanol

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

CC(C)(C)O[C@@H]1C[C@H](N(C1)C(=O)OCC2c3ccccc3-c4ccccc24)C(O)=O

InChI

1S/C24H27NO5/c1-24(2,3)30-15-12-21(22(26)27)25(13-15)23(28)29-14-20-18-10-6-4-8-16(18)17-9-5-7-11-19(17)20/h4-11,15,20-21H,12-14H2,1-3H3,(H,26,27)/t15-,21+/m1/s1

InChI key

WPBXBYOKQUEIDW-VFNWGFHPSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Eugene Rogozhin et al.
International journal of molecular sciences, 19(11) (2018-11-08)
Herein, we describe a modified form of the antimicrobial hairpin-like peptide EcAMP1, isolated from barnyard grass (E. crusgalli) seeds, which is structurally characterized by a combination of high-pressure liquid chromatography, mass spectrometry, and automated Edman sequencing. This derivate has a
Julien Echterbille et al.
Toxicon : official journal of the International Society on Toxinology, 130, 1-10 (2017-02-28)
Animal toxins are peptides that often bind with remarkable affinity and selectivity to membrane receptors such as nicotinic acetylcholine receptors (nAChRs). The latter are, for example, targeted by α-conotoxins, a family of peptide toxins produced by venomous cone snails. nAChRs
Lucas L Bennink et al.
Biomaterials, 183, 67-76 (2018-08-28)
Degradation of the extracellular matrix (ECM) is one of the fundamental factors contributing to a variety of life-threatening or disabling pathological conditions. However, a thorough understanding of the degradation mechanism and development of new ECM-targeting diagnostics are severely hindered by
Yang Li et al.
Macromolecular bioscience, 15(1), 52-62 (2014-12-06)
To address the downside of conventional photo-patterning which can alter the chemical composition of protein scaffolds, we developed a non-covalent photo-patterning strategy for gelatin (denatured collagen) hydrogels that utilizes UV activated triple helical hybridization of caged collagen mimetic peptide (caged

Articles

Proline analogues are promising candidates for tuning the biological, pharmaceutical, or physicochemical properties of naturally occuring, as well as de novo designed, linear, and, cyclic peptides.

Proline analogues are promising candidates for tuning the biological, pharmaceutical, or physicochemical properties of naturally occuring, as well as de novo designed, linear, and, cyclic peptides.

Proline analogues are promising candidates for tuning the biological, pharmaceutical, or physicochemical properties of naturally occuring, as well as de novo designed, linear, and, cyclic peptides.

Proline analogues are promising candidates for tuning the biological, pharmaceutical, or physicochemical properties of naturally occuring, as well as de novo designed, linear, and, cyclic peptides.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service