Skip to Content
Merck
All Photos(2)

Key Documents

472514

Sigma-Aldrich

2-Naphthyl isocyanate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7NCO
CAS Number:
Molecular Weight:
169.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

bp

83 °C/0.2 mmHg (lit.)

mp

53-56 °C (lit.)

SMILES string

O=C=Nc1ccc2ccccc2c1

InChI

1S/C11H7NO/c13-8-12-11-6-5-9-3-1-2-4-10(9)7-11/h1-7H

InChI key

XIXJQNFTNSQTBT-UHFFFAOYSA-N

General description

2-Naphthyl isocyanate is an aromatic isocyanate. It is formed during the rat liver microsome catalyzed oxidative metabolism of 2-naphthyl isothiocyanate.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Tamura et al.
Carcinogenesis, 11(11), 2009-2014 (1990-11-01)
Calf thymus DNA and M13mp9 RF DNA were modified with [ring-3H]2-naphthyl isocyanate (NIC) and analyzed by reverse-phase HPLC following enzymatic hydrolysis. In each case, essentially, a single radioactive component, which co-chromatographed with authentic N4-2-naphthyl-carbamoyl-2'-deoxycytidine (NCdC), was detected. In order to
M S Lee
Chemical research in toxicology, 5(6), 791-796 (1992-11-01)
The present study investigated the oxidative metabolism of 2-naphthyl isothiocyanate catalyzed by rat liver microsomes. Incubation of 2-naphthyl isothiocyanate, microsomes, and NADPH yielded either N,N'-di-2-naphthylurea or, on inclusion of 2-aminofluorene in the incubations, N-2-naphthyl-N'-2-fluorenylurea. These ureas were formed by the
Yu Zheng et al.
Nature communications, 5, 5772-5772 (2014-12-17)
The self-assembly of nanoscale materials to form hierarchically ordered structures promises new opportunities in drug delivery, as well as magnetic materials and devices. Herein, we report a simple means to promote the self-assembly of two polymers with functional groups at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service