Skip to Content
Merck
All Photos(1)

Key Documents

N4525

Sigma-Aldrich

1-Naphthyl isothiocyanate

95%

Synonym(s):

α-Naphthyl isothiocyanate, 1-Isothiocyanatonaphthalene, Naphthalen-1-yl isothiocyanate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7NCS
CAS Number:
Molecular Weight:
185.24
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

mp

55.5-57 °C (lit.)

SMILES string

S=C=Nc1cccc2ccccc12

InChI

1S/C11H7NS/c13-8-12-11-7-3-5-9-4-1-2-6-10(9)11/h1-7H

InChI key

JBDOSUUXMYMWQH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Naphthyl isothiocyanate (NITC) can be used as a source of thiourea moiety to synthesize derivatives of calix[4]arenes to be used as anion acceptors. It can also be used for the synthesis of naphthyl substituted 1,2,4-triazoles and 1,3,4-thiadiazoles.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and antimicrobial activity of some pyridyl and naphthyl substituted 1, 2, 4-triazole and 1, 3, 4-thiadiazole derivatives.
Zamani K, et al.
Turkish Journal of Chemistry, 28(1), 95-100 (2004)
Calix [4] arenes containing thiourea and amide moieties: neutral receptors towards ?, ?-dicarboxylate anions.
Liu SY, et al.
Organic & Biomolecular Chemistry, 2(11), 1582-1586 (2004)
Bo Kong et al.
Toxicology and applied pharmacology, 261(2), 189-195 (2012-04-24)
Emerging evidence suggests that feeding a high-fat diet (HFD) to rodents affects the expression of genes involved in drug transport. However, gender-specific effects of HFD on drug transport are not known. The multidrug resistance-associated protein 2 (Mrp2, Abcc2) is a
Keith A Thatch et al.
Journal of pediatric surgery, 45(1), 89-94 (2010-01-29)
Dependence on total parenteral nutrition in intestinal failure or short bowel syndrome patients can lead to many complications. The most significant complication is progressive liver injury leading to liver failure. This study assesses the potential of hepatocyte growth factor (HGF)
Hossain M Golbar et al.
Toxicologic pathology, 39(5), 795-808 (2011-07-20)
To investigate pathogenesis of post-bile duct (BD) injury fibrosis, interlobular BD epithelial injury was induced in male F344 rats by a single IP injection of α-naphthylisothiocyanate (75 mg/kg body weight) and rats were observed for 12 days. On days 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service