Skip to Content
Merck
All Photos(1)

Key Documents

447315

Sigma-Aldrich

Lauryl acrylate

technical grade, 90%

Synonym(s):

Dodecyl acrylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCOOC12H25
CAS Number:
Molecular Weight:
240.38
Beilstein:
1779069
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

grade

technical grade

Assay

90%

contains

60-100 ppm monomethyl ether hydroquinone as inhibitor

refractive index

n20/D 1.445 (lit.)

density

0.884 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCOC(=O)C=C

InChI

1S/C15H28O2/c1-3-5-6-7-8-9-10-11-12-13-14-17-15(16)4-2/h4H,2-3,5-14H2,1H3

InChI key

PBOSTUDLECTMNL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Lauryl acrylate monomers are extremely hydrophobic in nature.

Application

Lauryl acrylate can undergo atom transfer radical polymerization (ATRP) to form its polymer. Lauryl acrylate may undergo photoinitiated polymerization in the presence of N-acetyl-4-nitro-1-naphthylamine (ANNA) and N,N-dimethylaniline (DMA).

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Photoinitiated bulk polymerization of lauryl acrylate by N-acetyl-4-nitro-1-naphthylamine in the presence of N,N-dimethylaniline
Sastre R, et al.
Journal of Photochemistry and Photobiology A: Chemistry, 44(1), 111-122 (1988)
Grafting polyacrylates on natural rubber latex, by miniemulsion polymerization
Ragupathy L, et al.
Colloid and Polymer Science, 289, 229?235-229?235 (2011)
Shihong Deng et al.
Advanced materials (Deerfield Beach, Fla.), 31(39), e1903970-e1903970 (2019-08-14)
3D printing has witnessed a new era in which highly complexed customized products become reality. Realizing its ultimate potential requires simultaneous attainment of both printing speed and product versatility. Among various printing techniques, digital light processing (DLP) stands out in
The atom transfer radical polymerization of lauryl acrylate.
Beers KL and Matyjaszewski K
Journal of Macromolecular Science, Part A: Pure and Applied Chemistry, 38(7) (2001)
Fulvio Grigolato et al.
ACS nano, 11(11), 11358-11367 (2017-10-19)
Interactions between proteins and surfaces in combination with hydrodynamic flow and mechanical agitation can often trigger the conversion of soluble peptides and proteins into aggregates, including amyloid fibrils. Despite the extensive literature on the empirical effects of surfaces and mechanical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service