Skip to Content
Merck
All Photos(1)

Key Documents

411442

Sigma-Aldrich

Stearyl methacrylate

Mixture of stearyl and cetyl methacrylates, contains MEHQ as inhibitor

Synonym(s):

N-Octadecyl methacrylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=C(CH3)CO2(CH2)17CH3
CAS Number:
Molecular Weight:
338.57
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

contains

MEHQ as inhibitor

refractive index

n20/D 1.451 (lit.)

bp

195 °C/6 mmHg (lit.)

mp

18-20 °C (lit.)

density

0.864 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCCCOC(=O)C(C)=C

InChI

1S/C22H42O2/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-24-22(23)21(2)3/h2,4-20H2,1,3H3

InChI key

HMZGPNHSPWNGEP-UHFFFAOYSA-N

Application

  • Coconut Oil Encapsulated Microcapsules: Design and synthesis of poly(stearyl methacrylate-co-hydroxyethyl metacrylate) based microcapsules for use as phase change materials, encapsulating coconut oil for thermal energy storage (Oktay, Baştürk, Kahraman, 2019).
  • Biocompatible Multifunctional Hydrogels: Development of biocompatible hydrogels using stearyl methacrylate and vinylpyrroliDonefor potential medical applications such as tissue engineering (Kılıç, Tuncaboylu, Argun, 2022).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liping Liang et al.
Polymers, 11(12) (2019-12-18)
A polyurethane (PU) sponge with hydrophobic/oleophilic property was prepared based on the thiol-ene click reaction, which was a simple method with only one step. Photopolymerization was induced through UV light on the sponge surface in a homogeneous solution containing polyethylene
Jie Ding et al.
Carbohydrate polymers, 247, 116686-116686 (2020-08-25)
Conductive and self-healing hydrogel sensor is perspective in human-machine interaction applications. However, the design of ideal self-healing hydrogels are always challenging. Herein, by introducing disulfide modified Ag nanowires (AgNWs), we show a novel self-healing hydrogel strain sensor with superior mechanics
Thotsaphorn Chaloemsuwiwattanakan et al.
Journal of separation science, 39(18), 3521-3527 (2016-07-23)
A simple and rapid method based on micro-liquid chromatography using a synthetic monolithic capillary column was developed for determination of iohexol in human serums, a marker to evaluate the glomerular filtration rate. A hydrophilic methacrylic acid-ethylene dimethacrylate monolith provided excellent
Ju Fang et al.
Acta biomaterialia, 88, 503-513 (2019-02-18)
The design of hydrogels with adequate mechanical properties and excellent bioactivity, osteoconductivity, and capacity for osseointegration is essential to bone repair and regeneration. However, it is challenging to integrate all these properties into one bone scaffold. Herein, we developed a
Houchao Jing et al.
Soft matter, 15(26), 5264-5270 (2019-06-18)
Hydrogels with multiple shape-memory ability have aroused great interest due to their promising applications in various fields. Nevertheless, the weak mechanical performance of most shape-memory hydrogels seriously impedes the practical application in more complex environments. Herein, we reported a novel

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service