Skip to Content
Merck
All Photos(1)

Key Documents

437808

Sigma-Aldrich

(3-Bromopropyl)trichlorosilane

90%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br(CH2)3SiCl3
CAS Number:
Molecular Weight:
256.43
Beilstein:
1737852
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

90%

form

liquid

refractive index

n20/D 1.494 (lit.)

bp

202-204 °C (lit.)

density

1.605 g/mL at 25 °C (lit.)

functional group

bromo

SMILES string

Cl[Si](Cl)(Cl)CCCBr

InChI

1S/C3H6BrCl3Si/c4-2-1-3-8(5,6)7/h1-3H2

InChI key

UUNGBOQAZQUJMZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(3-Bromopropyl)trichlorosilane can be used:
  • To prepare coupling agent 3-iodopropyltrichlorosilane (3-IC3H6SiCl3), which is employed for the preparation of self-assembled nonlinear optically (NLO) active chromophoric multilayers.
  • As a reagent for the surface modification of silica nanofiber films and silica particles.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

168.8 °F

Flash Point(C)

76 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tris (hydroxymethyl) aminomethane?functionalized silica particles and their application for hydrophilic interaction chromatography.
Bui NTH, et al.
Journal of Separation Science, 33(19), 2965-2976 (2010)
Atomic resolution X-ray standing wave microstructural characterization of NLO-active self-assembled chromophoric superlattices.
Lin W, et al.
Journal of the American Chemical Society, 119(9), 2205-2211 (1997)
Bromopropyl functionalized silica nanofibers for effective removal of trace level dieldrin from water
Xiuli Y, et al.
Colloids and Surfaces. A, Physicochemical and Engineering Aspects, 406, 44-51 (2012)
Bromopropyl functionalized silica nanofibers for effective removal of trace level dieldrin from water
Yue X, et al.
Colloids and Surfaces. A, Physicochemical and Engineering Aspects, 406(9), 44-51 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service