Skip to Content
Merck
All Photos(2)

Key Documents

448931

Sigma-Aldrich

Trichloro(1H,1H,2H,2H-perfluorooctyl)silane

97%

Synonym(s):

Trichloro(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)silane, 1H,1H,2H,2H-Perfluorooctyl-trichlorosilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3(CF2)5CH2CH2SiCl3
CAS Number:
Molecular Weight:
481.54
Beilstein:
6154526
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

97%

refractive index

n20/D 1.352 (lit.)

bp

192 °C (lit.)

density

1.3 g/mL at 25 °C (lit.)

SMILES string

FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CC[Si](Cl)(Cl)Cl

InChI

1S/C8H4Cl3F13Si/c9-25(10,11)2-1-3(12,13)4(14,15)5(16,17)6(18,19)7(20,21)8(22,23)24/h1-2H2

InChI key

PISDRBMXQBSCIP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Trichloro(1H,1H,2H,2H-perfluorooctyl)silane (PFOCTS) is a chlorosilane that forms a self-assembled monolayer (SAM) on a variety of substrates. It is majorly used to modify the surface with superhydrophobic properties. It is a biocompatible polymer with low surface energy. PFOCTS exhibits anti-fouling properties with high wettability which facilitates the release of different composites from the mold.

Application

PFOCTS is a coupling agent that is used to silanize the substrate for the fabrication of microfluidic devices using soft lithography. It can also be used on silicon base that forms a chemically patterned surface for quantitative analysis of insoluble surfactants in the polymeric matrix. Alumina membranes can be treated with PFOCTS for the formation of surperhydrophobic polyimide (PI) based nanotube arrays.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biomimetic polyimide nanotube arrays with slippery or sticky superhydrophobicity
Zhu S, et al.
Journal of Colloid and Interface Science, 344(2), 541-546 (2010)
Extensive adipogenic and osteogenic differentiation of patterned human mesenchymal stem cells in a microfluidic device
Tenstad Ellen, et al.
Lab on a chip, 10(11), 1401-1409 (2010)
Soluble surfactant spreading on spatially confined thin liquid films
Hanyak M, et al.
Soft Matter, 8(29), 7660-7671 (2012)
Superhydrophobic surfaces fabricated by microstructuring of stainless steel using a femtosecond laser.
Wu B, et al.
Applied Surface Science, 256(1), 61-66 (2009)
Immobilization of polysaccharides on a fluorinated silicon surface
Wang A, et al.
Colloids and Surfaces. B, Biointerfaces, 47(1), 57-63 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service