Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Lithium perchlorate
Linear Formula:
LiClO
4
CAS Number:
7791-03-9
Molecular Weight:
106.39
EC Number:
232-237-2
Products
Product Category
salts (3)
electrolytes (1)
Brand
Sigma-Aldrich (7)
Sustainability
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Purity
Quality Segment
Application
Reagent Type
Special Grade
ACS reagent (1)
Purum (1)
Purum P.A. (1)
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
205281
Lithium perchlorate,
ACS reagent, ≥95.0%
Expand
View Pricing
634565
Lithium perchlorate,
battery grade, dry, 99.99% trace metals basis
Expand
View Pricing
431567
Lithium perchlorate,
99.99% trace metals basis
Expand
View Pricing
62580
Lithium perchlorate,
purum p.a., ≥98.0% (calc. based on dry substance, T), powder
Expand
View Pricing
931969
Lithium perchlorate,
anhydrous, ≥99.9% trace metals basis
Expand
View Pricing
936650
Lithium perchlorate,
Technipur
®
, battery grade, dry, 99.99% trace metals basis
Expand
View Pricing
18-1605
Lithium perchlorate,
SAJ special grade, ≥99.0%
Expand
View Pricing