Skip to Content
Merck
All Photos(1)

Documents

423270

Sigma-Aldrich

2-Thiopheneethylamine

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H9NS
CAS Number:
Molecular Weight:
127.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

liquid

refractive index

n20/D 1.551 (lit.)

bp

200-201 °C/750 mmHg (lit.)

density

1.087 g/mL at 25 °C (lit.)

SMILES string

NCCc1cccs1

InChI

1S/C6H9NS/c7-4-3-6-2-1-5-8-6/h1-2,5H,3-4,7H2

InChI key

HVLUYXIJZLDNIS-UHFFFAOYSA-N

General description

2-Thiopheneethylamine (2-(thiophen-2-yl)ethanamine) is an aromatic amine. It undergoes microwave induced condensation with iminodiacetic acid to form the corresponding piperazine-2,6-dione derivatives. Its effect as a probable substitute to the pyridine ligand on the performance of poly(3-hexylthiophene)/CdSe hybrid solar cells has been investigated.

Application

2-Thiopheneethylamine (2-thiophene ethyl amine, 2-(thien-2-yl)ethylamine) is suitable to functionalize multiwall carbon nanotubes (MWCNT).
It may be used as a reactant in the synthesis of pyrimidine derivatives by reacting with various isothiocyanatoketones and acylguanidines derivatives by reacting with aroyl S-methylisothiourea.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

190.4 °F - closed cup

Flash Point(C)

88 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

S Padmanabhan et al.
Bioorganic & medicinal chemistry letters, 11(24), 3151-3155 (2001-11-27)
Solution-phase synthesis of various acylguanidine derivatives and the evaluation of a small library of compounds as potential sodium channel blockers are described.
Synthesis, anti-inflammatory and analgesic activity evaluation of some pyrimidine derivatives.
Sondhi SM, et al.
Indian J. Chem. B, 48(2), 273-273 (2009)
Efficient synthesis of piperazine-2,6-dione and 4-(1H-indole-2-carbonyl) piperazine-2,6-dione derivatives and their evaluation for anticancer activity.
Kumar S, et al.
Medicinal Chemistry Research, 22(10), 4600-4609 (2013)
Veera Sadhu et al.
Nanotechnology, 22(26), 265607-265607 (2011-05-18)
We have successfully prepared mono- and bi-functionalized multiwall carbon nanotubes (MWCNT) with thiophene, amine and thiophene-amine groups. The dispersion of nanotubes has been enhanced and stable optimized dispersions in organic solvents were obtained. These functionalized nanotubes have been successfully incorporated
Lu Wang et al.
Materials science & engineering. C, Materials for biological applications, 88, 157-165 (2018-04-11)
Hyaluronic acid (HA) is a component of extracellular matrix, which is important for cell functions and tissue integrity. Biosynthesized HA, as well as its derivatives, is widely used in cosmetics industry, biochemical medicine and medical surgery. In this research, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service