Skip to Content
Merck
All Photos(1)

Key Documents

416487

Sigma-Aldrich

2-Octen-1-ylsuccinic anhydride, mixture of cis and trans

97%

Synonym(s):

2-Octenylsuccinic anhydride, 3-(Oct-2-enyl)dihydrofuran-2,5-dione, Dihydro-3-(2-octen-1-yl)-2,5-furandione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H18O3
CAS Number:
Molecular Weight:
210.27
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

liquid

refractive index

n20/D 1.4694 (lit.)

bp

168 °C/10 mmHg (lit.)

mp

8-12 °C (lit.)

density

1 g/mL at 25 °C (lit.)

SMILES string

CCCCC\C=C\CC1CC(=O)OC1=O

InChI

1S/C12H18O3/c1-2-3-4-5-6-7-8-10-9-11(13)15-12(10)14/h6-7,10H,2-5,8-9H2,1H3/b7-6+

InChI key

WSGFXVFLWVXTCJ-VOTSOKGWSA-N

Application

  • Biosynthesis of pectic oligosaccharide-based amphiphiles as novel stabilizers of nanoemulsions by coupling enzymatic depolymerization with alkyl/alkenyl: This study explores the physicochemical properties of novel stabilizers, including 2-Octen-1-ylsuccinic anhydride, for use in food hydrocolloids (L Li et al., 2023).
  • Enzymatic alkylsuccinylation of tyrosol: Synthesis, characterization and property evaluation as a dual-functional antioxidant: Details the synthesis of derivatives of tyrosol using 2-Octen-1-ylsuccinic anhydride, assessing their antioxidant properties (S Marzocchi et al., 2018).
  • Dynamic high-pressure microfluidization assisting octenyl succinic anhydride modification of rice starch: Describes the use of dynamic high-pressure techniques to modify rice starch with 2-Octen-1-ylsuccinic anhydride for improved performance in food applications (Y Li et al., 2018).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Victor D Quintero-Castaño et al.
Food chemistry, 315, 126241-126241 (2020-02-06)
Chemical modification with octenyl succinic anhydride (OSA) helps to control the physicochemical and thermal properties of isolated starches. The main objective, herein, was to partially characterize modified starches from Dominico-Harton plantain and FHIA 21 planted in Colombia. The highest degree
Yunia Verónica García-Tejeda et al.
Journal of food science and technology, 55(6), 2279-2287 (2018-06-13)
The encapsulation by spray drying of maize anthocyanins was evaluated using two types of wall materials, consisting of normal and waxy maize starch, which were esterified with octenyl succinic anhydride. The X-ray diffraction analysis revealed that SWMS possessed a completely

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service