Skip to Content
Merck
All Photos(1)

Key Documents

216445

Supelco

Amberlite IRA743 Ion Exchange Resin

free base, 25-30 mesh

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
23151817
NACRES:
SB.52

Product Name

Amberlite IRA743 free base, free base

vapor density

<1 (vs air)

Quality Level

vapor pressure

17 mmHg ( 20 °C)

description

weakly basic anion exchange resin

form

beads

autoignition temp.

~800 °F

moisture

48-54%

technique(s)

LPLC: suitable

matrix

styrene-divinylbenzene (macroporous)

matrix active group

N-methylglucamine (free base form) functional group

particle size

500-700 μm

capacity

0.7 meq/mL by wetted bed volume

separation technique

affinity

SMILES string

N(C(O)[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO)(Cc2c(cccc2)C=C)C.c1(c(cccc1)C=C)C=C

InChI

1S/C16H25NO6.C10H10/c1-3-10-6-4-5-7-11(10)8-17(2)16(23)15(22)14(21)13(20)12(19)9-18;1-3-9-7-5-6-8-10(9)4-2/h3-7,12-16,18-23H,1,8-9H2,2H3;3-8H,1-2H2/t12-,13-,14+,15-,16?;/m1./s1

InChI key

STVMLWQMXHXTJS-ULVCUCBLSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Amberlite IRA743 free base is of styrenic, macroreticular form.

Application

Borate-specific chelating resin for the removal of borate, boric acid, other boron species from water; highly selective so that salts, including bases, do not interfere significantly.

Features and Benefits

Weakly basic resin with N-methylglucaomine functionality which is useful in boron-specific applications.

Legal Information

Amberlite is a trademark of DuPont de Nemours, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adsorption mechanism of hexavalent chromium removal using Amberlite IRA 743 resin.
Gandhi
Ion Exchange Letters , 3, 25-35 (2010)
Boron pollution in the Simav River, Turkey and various methods of boron removal.
Okay, O., et al.
Water Research, 19, 857-862 (1985)
Lei Zhang et al.
Nature communications, 11(1), 5371-5371 (2020-10-25)
Autoinducer-2 (AI-2) is a quorum sensing signal that mediates communication within and between many bacterial species. However, its known receptors (LuxP and LsrB families) are not found in all the bacteria capable of responding to this signaling molecule. Here, we
Catalysis by Anion Exchange Resins.
Schmidle, Claude J., and Richard C. Mansfield.
Ind. and Eng. Chem., 44, 1388-1390 (1952)

Related Content

Research spans transition-metal catalysis, boron chemistry, organocatalysis, multicomponent reactions, and sustainable methods.

Research spans transition-metal catalysis, boron chemistry, organocatalysis, multicomponent reactions, and sustainable methods.

Research spans transition-metal catalysis, boron chemistry, organocatalysis, multicomponent reactions, and sustainable methods.

Research spans transition-metal catalysis, boron chemistry, organocatalysis, multicomponent reactions, and sustainable methods.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service