Skip to Content
Merck
All Photos(2)

Key Documents

413631

Sigma-Aldrich

Calcium bromide

AnhydroBeads, −10 mesh, 99.98% trace metals basis

Synonym(s):

Hydrobromic acid calcium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaBr2
CAS Number:
Molecular Weight:
199.89
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

product line

AnhydroBeads

Assay

99.98% trace metals basis

form

beads

impurities

≤250.0 ppm Trace Metal Analysis

particle size

−10 mesh

density

3.353 g/mL at 25 °C (lit.)

SMILES string

Br[Ca]Br

InChI

1S/2BrH.Ca/h2*1H;/q;;+2/p-2

InChI key

WGEFECGEFUFIQW-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

AnhydroBeads is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Johnson Wright et al.
Journal of contaminant hydrology, 115(1-4), 46-63 (2010-05-07)
The behavior of dense, viscous calcium bromide brine solutions used to remediate systems contaminated with dense nonaqueous phase liquids (DNAPLs) is considered in laboratory and field porous medium systems. The density and viscosity of brine solutions are experimentally investigated and
Curtis D Mowry et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 32(2), 177-182 (2016-02-11)
Trace moisture quantitation is crucial in medical, civilian and military applications. Current aquametry technologies are limited by the sample volume, reactivity, or interferences, and/or instrument size, weight, power, cost, and complexity. We report for the first time on the use
Katarzyna Rlepokura et al.
Acta crystallographica. Section C, Crystal structure communications, 64(Pt 3), m127-m133 (2008-03-07)
Two hydrated complexes of monomeric dihydroxyacetone (DHA; the simplest ketose), viz. the calcium bromide complex bis(mu-dihydroxyacetone)bis[tetraaquacalcium(II)] tetrabromide (isomorphous with the chloride compound reported previously), [Ca(2)(C(3)H(6)O(3))(2)(H(2)O)(8)]Br(4), (2e), and the cadmium chloride complex poly[[bis(mu-dihydroxyacetone)bis[bis(dihydroxyacetone)cadmium(II)]] [diaquatetradeca-mu-chlorido-dichloridohexacadmium(II)] tetrahydrate], {[Cd(2)(C(3)H(6)O(3))(6)][Cd(6)Cl(16)(H(2)O)(2)] x 4 H(2)O}(n), (2f), are
Kathleen D Carillo et al.
International journal of molecular sciences, 21(17) (2020-09-02)
Gramicidin A (gA) forms several convertible conformations in different environments. In this study, we investigated the effect of calcium halides on the molecular state and antimicrobial activity of gramicidin A. The molecular state of gramicidin A is highly affected by
Shinji Sasada et al.
International journal of clinical oncology, 12(4), 274-278 (2007-08-19)
Paclitaxel often causes severe hypersensitivity reactions (HSRs) rapidly after infusion, even in patients given prophylactic therapy. The purpose of this study was to analyze the incidence of paclitaxel-related HSRs in patients with non-small cell lung cancer (NSCLC) retrospectively, and to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service