Skip to Content
Merck
All Photos(3)

Documents

310034

Sigma-Aldrich

Calcium carbonate

powder, ≤50 μm particle size, 98%

Synonym(s):

Calcite, Calcium monocarbonate, Monocalcium carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaCO3
CAS Number:
Molecular Weight:
100.09
Beilstein:
8008338
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

98%

form

powder

particle size

≤50 μm

solubility

H2O: insoluble

density

2.93 g/mL at 25 °C (lit.)

SMILES string

[Ca++].[O-]C([O-])=O

InChI

1S/CH2O3.Ca/c2-1(3)4;/h(H2,2,3,4);/q;+2/p-2

InChI key

VTYYLEPIZMXCLO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Calcium Carbonate (CaCO3) is a naturally found material in chalk, limestone, and marble. It is composed of three elements which include carbon, oxygen, and calcium. It is formed by reacting carbon dioxide with slaked or burnt lime. It can be used for a variety of applications ranging from industrial, food to agriculture.

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Calcium carbonate: from the Cretaceous period into the 21st century (2001)
Sophie A Jamal et al.
Lancet (London, England), 382(9900), 1268-1277 (2013-07-23)
Phosphate binders (calcium-based and calcium-free) are recommended to lower serum phosphate and prevent hyperphosphataemia in patients with chronic kidney disease, but their effects on mortality and cardiovascular outcomes are unknown. We aimed to update our meta-analysis on the effect of
Polysaccharides and proteoglycans in calcium carbonate-based biomineralization.
José L Arias et al.
Chemical reviews, 108(11), 4475-4482 (2008-07-26)
M Azizur Rahman et al.
PloS one, 8(3), e58781-e58781 (2013-03-26)
An organic matrix consisting of a protein-polysaccharide complex is generally accepted as an important medium for the calcification process. While the role this "calcified organic matrix" plays in the calcification process has long been appreciated, the complex mixture of proteins
Jane A Cauley et al.
Journal of women's health (2002), 22(11), 915-929 (2013-10-18)
Clinical outcomes of the Women's Health Initiative (WHI) calcium plus vitamin D supplementation trial have been reported during 7.0 years of active intervention. We now report outcomes 4.9 years after the intervention stopped and cumulative findings. Postmenopausal women (N=36,282) were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service