Skip to Content
Merck
All Photos(1)

Documents

411736

Sigma-Aldrich

1,6-Hexanediol dimethacrylate

contains 75.0-125.0 hydroquinone as inhibitor

Synonym(s):

1,6-Hexamethylene dimethacrylate, 1,6-Hexanediyl dimethacrylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[H2C=C(CH3)CO2(CH2)3-]2
CAS Number:
Molecular Weight:
254.32
Beilstein:
1957290
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

0.02 mmHg ( 100 °C)

Quality Level

Assay

≥90%

form

liquid

contains

75.0-125.0 hydroquinone as inhibitor

refractive index

n20/D 1.458 (lit.)

bp

>315 °C (lit.)

density

0.995 g/mL at 25 °C (lit.)

SMILES string

CC(=C)C(=O)OCCCCCCOC(=O)C(C)=C

InChI

1S/C14H22O4/c1-11(2)13(15)17-9-7-5-6-8-10-18-14(16)12(3)4/h1,3,5-10H2,2,4H3

InChI key

SAPGBCWOQLHKKZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D R Williams et al.
Primary dental care : journal of the Faculty of General Dental Practitioners (UK), 8(4), 151-155 (2002-01-22)
The fit of the denture base to the alveolar ridge progressively declines with alveolar ridge resorption. Once a denture becomes ill-fitting there are two main treatment options: to construct a new denture or to restore the fit of the existing
N M Mohsen et al.
Journal of oral rehabilitation, 22(3), 183-189 (1995-03-01)
The effect of silanation on the dispersion of quartz and zirconia-silica fillers by monomers used to formulate composites was studied. Sixteen silane coupling agents and three methods were used, and the amounts of the monomers needed to reach wet and
M Yatabe et al.
Journal of oral rehabilitation, 28(2), 180-185 (2001-04-12)
The purpose of this study was to clarify the effect of the reducing agent on the oxygen-inhibited layer of the cross-linked reline material. A commercial autopolymerizing reline resin containing 1,6-hexanediol dimethacrylate as cross-linking agent and 1 wt.% sodium sulphite solution
C Park et al.
Dental materials : official publication of the Academy of Dental Materials, 14(6), 385-393 (1999-09-14)
To explore possible enhancement of the mechanical properties of resin composites by aligning the filler particles. The resin for the composites consisted of urethane dimethacrylate (UDMA) and 1,6-hexanediol dimethacrylate (HDDMA) mixed in the ratio of 90 to 10; the filler
William M Chirdon et al.
Dental materials : official publication of the Academy of Dental Materials, 22(1), 57-62 (2005-09-13)
This study aims to characterize the translucency of aligned, short-fiber composites by determining the effects of filler particle orientation on the Kubelka-Munk absorption and scattering coefficients. Rectangular composite specimens (n = 3) were filled with very short E-glass fibers. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service