Skip to Content
Merck
All Photos(1)

Key Documents

409456

Sigma-Aldrich

Tetrahydrofurfuryl methacrylate

contains 75 ppm HQ as inhibitor, 900 ppm MEHQ as inhibitor, 97%

Synonym(s):

(Tetrahydrofuran-2-yl)methyl methacrylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H14O3
CAS Number:
Molecular Weight:
170.21
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

97%

form

liquid

contains

75 ppm HQ as inhibitor
900 ppm MEHQ as inhibitor

refractive index

n20/D 1.458 (lit.)

bp

52 °C/0.4 mmHg (lit.)

density

1.044 g/mL at 25 °C (lit.)

SMILES string

CC(=C)C(=O)OCC1CCCO1

InChI

1S/C9H14O3/c1-7(2)9(10)12-6-8-4-3-5-11-8/h8H,1,3-6H2,2H3

InChI key

LCXXNKZQVOXMEH-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

195.8 °F - closed cup

Flash Point(C)

91 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M P Patel et al.
Biomaterials, 19(21), 1911-1917 (1998-12-24)
The release of fluoride ions from two room-temperature polymerising systems containing sodium and potassium fluoride, respectively, has been studied. The polymer systems comprised poly(ethyl methacrylate) powder (PEM), with tetrahydrofurfuryl methacrylate (THFM), and n-butyl methacrylate (nBM), respectively. The water uptake of
J J A Barry et al.
Biomaterials, 25(17), 3559-3568 (2004-03-17)
This paper describes a method of foaming a polymer system comprising poly(ethyl methacrylate)/tetrahydrofurfuryl methacrylate (PEMA/THFMA), characterisation of the resulting porosity and use of the foam for chondrocyte culture. The potential for this polymer system to support cartilage repair has been
M P Patel et al.
Biomaterials, 22(17), 2319-2324 (2001-08-21)
Oral candidal infections are often persistent and intractable and thus the aim of this study was to develop a polymeric sustained release device to improve the topical treatment of these infections. A self curing system based on poly(ethyl methacrylate) and
R Labella et al.
Biomaterials, 15(15), 1197-1200 (1994-12-01)
Newly designed composite resins for dental applications have been studied. The matrix phases were based on 2,2 bis-4(2-hydroxy-3-methacryloyloxypropoxy)phenyl propane and a urethane dimethacrylate. Tetrahydrofurfuryl methacrylate was added as a diluent to both dimethacrylates in a proportion of 30 wt%. Heat-cured
P D Riggs et al.
Biomaterials, 21(4), 345-351 (2000-02-03)
A series of different methacrylate monomers (with either 1 or 2.5% dimethyl-p-toluidine, DMPT) was gelled with poly(ethyl methacrylate) powder (containing benzoyl peroxide) thus forming a room temperature curing system. When doped with 5.625% chlorhexidine diacetate the release from the tetrahydrofurfuryl

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service