Skip to Content
Merck
All Photos(2)

Key Documents

409324

Sigma-Aldrich

Aluminum iodide

anhydrous, powder, 99.999% trace metals basis

Synonym(s):

Aluminum triiodide, Aluminum(III) iodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
AlI3
CAS Number:
Molecular Weight:
407.70
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.999% trace metals basis
grade:
anhydrous
form:
powder

grade

anhydrous

Quality Level

vapor pressure

1 mmHg ( 178 °C)

Assay

99.999% trace metals basis

form

powder

impurities

≤15.0 ppm Trace Metal Analysis

bp

360 °C (lit.)

mp

191 °C (lit.)

density

3.98 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

I[Al](I)I

InChI

1S/Al.3HI/h;3*1H/q+3;;;/p-3

InChI key

CECABOMBVQNBEC-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

General description

Aluminum iodide (AlI3) is a strong Lewis acid that is used as an electrolyte additive in electrochemical energy storage and conversion devices.

Application

Aluminum iodide can be used as:
  • An electrolyte additive for the pretreatment of Li metal electrodes. The addition of AlI3 provides a durable Li interface with high Li cyclability.
  • A homogeneous redox mediator for rechargeable Al-CO2batteries with high discharge voltage and a high capacity. These secondary batteries are promising rechargeable systems for future grid energy storage applications.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cheap and Environmentally Benign Electrochemical Energy Storage and Conversion Devices Based on AlI3 Electrolytes
Bofei Xue, et al.
Journal of the American Chemical Society, 128, 8720-8721 (2006)
Pretreatment of Li metal anode with electrolyte additive for enhancing Li cycleability
Masashi Ishikawa, et al.
Journal of Power Sources, 146, 199-203 (2005)
A Secondary Al?CO2 Battery Enabled by Aluminum Iodide as a Homogeneous Redox Mediator
Christopher Fetrow, et al.
ACS Applied Materials & Interfaces, 15, 12908-12914 (2023)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service