Skip to Content
Merck
All Photos(1)

Key Documents

383260

Sigma-Aldrich

Germanium(II) iodide

≥99.8% trace metals basis

Synonym(s):

Germanium diiodide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
GeI2
CAS Number:
Molecular Weight:
326.45
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99.8% trace metals basis

form

solid

reaction suitability

reagent type: catalyst
core: germanium

impurities

≤2000.0 ppm Trace Metal Analysis

density

5.37 g/mL at 25 °C (lit.)

SMILES string

I[Ge]I

InChI

1S/GeI2/c2-1-3

InChI key

IAGYEMVJHPEPGE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reagent forms stable ionic adducts with carbenes.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sara R Smock et al.
Nanoscale, 12(4), 2764-2772 (2020-01-21)
As surface ligands play a critical role in the colloidal stability and optoelectronic properties of semiconductor nanocrystals, we used solution NMR experiments to investigate the surface coordination chemistry of Ge nanocrystals synthesized by a microwave-assisted reduction of GeI2 in oleylamine.
Xiaotong Wu et al.
Chemistry, an Asian journal, 13(13), 1654-1659 (2018-04-28)
Colloidal nanocrystals (NCs) of metal halide perovskite have recently aroused great research interest, due to their remarkable optical and electronic properties. We report a solution synthesis of a new member in this category, that is, all-inorganic lead-free cesium germanium iodine
Inorganic Chemistry, 32, 1541-1541 (1993)
Kohei Nishimura et al.
ACS applied materials & interfaces, 11(34), 31105-31110 (2019-08-07)
In the composition of Q0.1(FA0.75MA0.25)0.9SnI3, Q is replaced with Na+, K+, Cs+, ethylammonium+ (EA+), and butylammonium+ (BA+), respectively, and the relationship between actually measured lattice strain and photovoltaic performances is discussed. The lattice strain evaluated by the Williamson-hall plot of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service