Skip to Content
Merck
All Photos(1)

Key Documents

408085

Sigma-Aldrich

3-Bromo-5-iodobenzoic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H3(I)CO2H
CAS Number:
Molecular Weight:
326.91
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

219-221 °C (lit.)

SMILES string

OC(=O)c1cc(Br)cc(I)c1

InChI

1S/C7H4BrIO2/c8-5-1-4(7(10)11)2-6(9)3-5/h1-3H,(H,10,11)

InChI key

MKJBJYCBKXPQSY-UHFFFAOYSA-N

General description

3-Bromo-5-iodobenzoic acid (BrIBA) is a halogen substituted carboxylic acid.

Application

3-Bromo-5-iodobenzoic acid may be used in the preparation of the following:
  • Phenyl(3-bromo-5-iodo)benzoate.
  • As starting reagent for the large-scale synthesis of the thromboxane receptor antagonist 3-{3-[2-(4-chlorobenzenesulfonamido)ethyl]-5-(4-fluorobenzyl)phenyl}propionic acid, via regioselective Heck cross-coupling reaction.
  • Methyl 3-bromo-5-iodobenzoate.
  • 3-Bromo-5-(triisopropylsilylethynyl)benzoic acid, via Sonogashira coupling reaction.
  • Trifluoroacetophenone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of acetylene-functionalized [2]rotaxane monomers directed toward side chain-type polyrotaxanes.
Nakazono K, et al.
Polymer Journal, 42(3), 208-215 (2010)
A scalable synthesis of the thromboxane receptor antagonist 3-{3-[2-(4-chlorobenzenesulfonamido) ethyl]-5-(4-fluorobenzyl) phenyl} propionic acid via a regioselective Heck cross-coupling strategy.
Waite DC and Mason CP.
Organic Process Research & Development, 2(2), 116-120 (1998)
S Lindman et al.
Bioorganic & medicinal chemistry, 8(9), 2375-2383 (2000-10-12)
Rigidification of peptides by cyclization and iterative incorporation of well-defined secondary structure mimetics constitutes one approach to the design of non-peptidergic structures with better defined conformations. We herein present the synthesis of a potential gamma-turn mimetic scaffold, and its incorporation
Keisuke Gondo et al.
Molecules (Basel, Switzerland), 17(6), 6625-6632 (2012-06-26)
Reaction of dibenzoylmethane with (diacetoxyiodo)benzene in the presence of KOH in MeCN quantitatively gave the corresponding iodonium ylide, which was treated with a HF reagent to afford the corresponding 2-fluorinated dibenzoylmethane in 14-50% yields. The similar reaction of the iodonium

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service