Skip to Content
Merck
All Photos(1)

Key Documents

407704

Sigma-Aldrich

4-Methylmorpholine

purified by redistillation, ≥99.5%

Synonym(s):

NMM

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H11NO
CAS Number:
Molecular Weight:
101.15
Beilstein:
102719
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
≥99.5%
bp:
115-116 °C/750 mmHg (lit.)
vapor pressure:
18 mmHg ( 20 °C)
Pricing and availability is not currently available.

vapor density

>1 (vs air)

Quality Level

vapor pressure

18 mmHg ( 20 °C)

Assay

≥99.5%

form

liquid

purified by

glass distillation
redistillation

refractive index

n20/D 1.435 (lit.)

pH

10.6 (20 °C, 50 g/L)

bp

115-116 °C/750 mmHg (lit.)

mp

−66 °C (lit.)

density

0.92 g/mL at 25 °C (lit.)

functional group

ether

SMILES string

CN1CCOCC1

InChI

1S/C5H11NO/c1-6-2-4-7-5-3-6/h2-5H2,1H3

InChI key

SJRJJKPEHAURKC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Methylmorpholine is an economical cation useful for the preparation of morpholinium cation-based ionic liquids.[1] The experimental FT-IR and FT-Raman spectra of 4-methylmorpholine has been recorded over the range 4000-400cm-1 and 3500-100cm-1 respectively.[2] It undergoes fluorination with cobalt(III) fluoride to afford highly fluorinated morpholines, which were characterized by mass and NMR spectroscopy.[3]

Application

4-Methylmorpholine may be used in the preparation of N-ethyl-N-methylmorpholinium bis(trifluoromethanesulfonyl)imide.[1]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

53.6 °F - closed cup - DIN 51755 Part 1

Flash Point(C)

12 °C - closed cup - DIN 51755 Part 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The fluorination of 4-methylmorpholine over cobalt (III) fluoride: The isolation and characterisation of some novel polyfluoro-4-methylmorpholines by mass spectrometry and nmr spectroscopy; the mechanism of the title reaction.
Rendell RW and Wright B.
Tetrahedron, 34(2), 197-203 (1978)
Ki-Sub Kim et al.
Chemical communications (Cambridge, England), (7)(7), 828-829 (2004-03-27)
Ionic liquids based on N-alkyl-N-methylmorpholinium salts have been synthesized and the physical and electrochemical characteristics of this family of ionic liquids have been investigated for use as electrolytes.
V Balachandran et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 97, 1101-1110 (2012-08-30)
The experimental FT-IR (4000-400 cm(-1)) and FT-Raman (3500-100 cm(-1)) spectra of 4-Methylmorpholine were recorded. The observed bands were interpreted with the aid of normal coordinate analysis and force field calculations based on density functional theory (DFT) using B3LYP functional theory
L Hansén et al.
Scandinavian journal of work, environment & health, 12(1), 66-69 (1986-02-01)
The N-methylmorpholine levels in workroom air in a polyurethane foam factory were determined by two methods in which midget impinger flasks were used for sampling. The analyses were performed by gas chromatography and isotachophoresis. The values obtained by the gas
K Sitarek
Teratogenesis, carcinogenesis, and mutagenesis, 19(6), 369-376 (1999-12-10)
N-methylmorpholine, which is used as a catalyst in polyurethane foams producing, in solvents, stabilizing agents, and corrosion inhibitors, was administered to female rats by gavage at 100, 200, 600, and 900 mg/kg during organogenesis. It did not exhibit selective toxicity

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service