Merck
DFT, FT-Raman, FT-IR, HOMO-LUMO and NBO studies of 4-Methylmorpholine.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2012-08-30)
V Balachandran, G Mahalakshmi, A Lakshmi, A Janaki
ABSTRACT
The experimental FT-IR (4000-400 cm(-1)) and FT-Raman (3500-100 cm(-1)) spectra of 4-Methylmorpholine were recorded. The observed bands were interpreted with the aid of normal coordinate analysis and force field calculations based on density functional theory (DFT) using B3LYP functional theory (DFT) using B3LYP functional with 6-311+G and 6-3++G basis sets. The complete assignments were performed on the basis of the potential energy distribution (PED) of the vibrational modes, calculated with scaled quantum mechanical (SQM) method. The molecular structure and vibrational frequencies, infrared intensities and Raman scattering actives have been calculated frequency showed the best agreement with experimental results. The formation of the hydrogen bond was investigated using NBO calculations. The calculated HOMO and LUMO energies show that charge transfer occur within the molecule. The dipolemoment (μ) and polarizability (α), anisotropy polarizability (Δα) and first hyperpolarizability (β(total)) of the molecule have been reported.
Sigma-Aldrich
4-Methylmorpholine, purified by redistillation, ≥99.5%
Sigma-Aldrich
4-Methylmorpholine, purum, ≥98.0% (GC)
Sigma-Aldrich
4-Methylmorpholine, BioXtra, suitable for protein sequencing, ≥99.5% (GC)
Sigma-Aldrich
4-Methylmorpholine, ReagentPlus®, 99%