Skip to Content
Merck
All Photos(1)

Documents

406147

Sigma-Aldrich

2,2′-Isopropylidenebis[(4S)-4-tert-butyl-2-oxazoline]

99%

Synonym(s):

(S,S)-2,2′-Isopropylidene-bis(4-tert-butyl-2-oxazoline)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H30N2O2
CAS Number:
Molecular Weight:
294.43
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

optical activity

[α]20/D −120°, c = 5 in chloroform

mp

89-91 °C (lit.)

SMILES string

CC(C)(C)[C@H]1COC(=N1)C(C)(C)C2=N[C@H](CO2)C(C)(C)C

InChI

1S/C17H30N2O2/c1-15(2,3)11-9-20-13(18-11)17(7,8)14-19-12(10-21-14)16(4,5)6/h11-12H,9-10H2,1-8H3/t11-,12-/m1/s1

InChI key

DPMGLJUMNRDNMX-VXGBXAGGSA-N

Application

Chiral catalyst for asymmetric synthesis. C2 symmetric ligand for enantioselective catalysis. Easily forms bidentate coordination complexes due to the strong affinity of the oxazoline nitrogen for various metals.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 44, 7849-7850 (2003)
L C Madoff et al.
Infection and immunity, 60(12), 4989-4994 (1992-12-01)
Group B streptococci (GBS) cause the majority of cases of neonatal sepsis and meningitis in the United States. Immunization of women of childbearing age is one strategy under consideration for the prevention of neonatal disease. The beta C protein, a
Nicholas Gathergood et al.
Organic & biomolecular chemistry, 2(7), 1077-1085 (2004-03-23)
The direct aldol reaction of 2-ketoesters catalyzed by chiral bisoxazoline copper(II) complexes has been investigated. First the direct homo-aldol reaction of ethyl pyruvate is reported which proceeds to give diethyl 2-hydroxy-2-methyl-4-oxoglutarate. This was isolated as the more stable optically active
Tetrahedron Letters, 45, 4213-4216 (2004)
Mauro Marigo et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 10(9), 2133-2137 (2004-04-28)
The first general catalytic asymmetric bromination and chlorination of beta-ketoesters has been developed. The reactions proceed for both acyclic and cyclic beta-ketoesters catalyzed by chiral bisoxazolinecopper(II) complexes giving the corresponding optically active alpha-bromo- and alpha-chloro-beta-ketoesters in high yields and moderate

Articles

BOX ligand complex enhances catalytic enantioselective aziridination of styrene, with phenyl substituents proving superior over t-Bu groups.

BOX ligand complex enhances catalytic enantioselective aziridination of styrene, with phenyl substituents proving superior over t-Bu groups.

BOX ligand complex enhances catalytic enantioselective aziridination of styrene, with phenyl substituents proving superior over t-Bu groups.

BOX ligand complex enhances catalytic enantioselective aziridination of styrene, with phenyl substituents proving superior over t-Bu groups.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service