Skip to Content
Merck
All Photos(3)

Key Documents

407240

Sigma-Aldrich

Copper(I) trifluoromethanesulfonate benzene complex

technical grade, 90%

Synonym(s):

Copper(I) triflate benzene complex, Cuprous trifluoromethanesulfonate benzene complex, Trifluoromethanesulfonic acid copper(I) salt benzene complex

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF3SO3Cu)2 · C6H6
CAS Number:
Molecular Weight:
503.34
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Quality Level

Assay

90%

form

powder

reaction suitability

core: copper
reagent type: catalyst

mp

160 °C (dec.) (lit.)

SMILES string

[Cu+].[Cu+].c1ccccc1.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F

InChI

1S/C6H6.2CHF3O3S.2Cu/c1-2-4-6-5-3-1;2*2-1(3,4)8(5,6)7;;/h1-6H;2*(H,5,6,7);;/q;;;2*+1/p-2

InChI key

GNXZWVVAAMVOJY-UHFFFAOYSA-L

Application

Copper(I) trifluoromethanesulfonate benzene complex can be used as a catalyst:
  • To synthesize enol-esters via copper(I) carboxylate intermediate formation.
  • In the enantioselective allylic oxidation of cyclic alkenes.
  • To prepare 2,5-disubstituted pyrrolidine derivatives from N-alkenyl, alkynyl and alkyl N-benzoyloxysulfonamides via the sulfonamidyl radical formation.

It can also be used in combination with amino acid-based chiral phosphine ligands to catalyze asymmetric conjugate additions of alkylzincs to acyclic α,β-unsaturated ketones, affording β-alkylcarbonyls in high yield and with excellent enantioselectivity.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Sol. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enantioselective total synthesis of erogorgiaene: applications of asymmetric Cu-catalyzed conjugate additions of alkylzincs to acyclic enones
Cesati RR, et al.
Journal of the American Chemical Society, 126(1), 96-101 (2004)
Synthesis of Enol Esters from Copper (I) Carboxylates Generated from Copper (I) Trifluoromethanesulfonate Benzene Complex
Lefler SR and Rose SD
Synthetic Communications, 29(21), 3805-3810 (1999)
Jerome Bayardon et al.
The Journal of organic chemistry, 69(9), 3121-3128 (2004-04-24)
Various enantiopure fluorous bis(oxazolines) with fluorine content between 52.7 and 58.7% have been synthesized by a simple reaction sequence that involved the introduction of two fluorinated ponytails by alkylation of the corresponding nonfluorous bis(oxazolines). These new ligands have been used
Enantiopure fluorous bis (oxazolines): Synthesis and applications in catalytic asymmetric reactions
Bayardon J and Sinou D
The Journal of Organic Chemistry, 69(9), 3121-3128 (2004)
Richard R Cesati et al.
Journal of the American Chemical Society, 126(1), 96-101 (2004-01-08)
The first enantioselective synthesis of erogorgiaene (1), an inhibitor of mycobacterium tuberculosis, is disclosed. The total synthesis highlights the utility of asymmetric conjugate additions (ACA) of alkylzincs to acyclic alpha,beta-unsaturated ketones catalyzed by peptidic phosphine ligands and (CuOTf)(2).C(6)H(6). Moreover, several

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service