Skip to Content
Merck
All Photos(3)

Key Documents

108987

Sigma-Aldrich

Methyl 3-mercaptopropionate

98%

Synonym(s):

3-Mercaptopropionic Acid Methyl Ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2CH2COOCH3
CAS Number:
Molecular Weight:
120.17
Beilstein:
1742952
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

refractive index

n20/D 1.464 (lit.)

bp

54-55 °C/14 mmHg (lit.)

density

1.085 g/mL at 25 °C (lit.)

SMILES string

COC(=O)CCS

InChI

1S/C4H8O2S/c1-6-4(5)2-3-7/h7H,2-3H2,1H3

InChI key

LDTLDBDUBGAEDT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

152.6 °F - closed cup

Flash Point(C)

67 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sungyoup Hong et al.
PloS one, 12(1), e0169262-e0169262 (2017-01-07)
In this report, we demonstrate the pH-dependent, in vitro antimicrobial activity of a cationic, amphiphilic random copolymer against clinical isolates of drug-resistant Staphylococcus aureus. The polymer was developed toward a long-term goal of potential utility in the treatment of skin
Cheng-Ju Yu et al.
Langmuir : the ACS journal of surfaces and colloids, 24(21), 12717-12722 (2008-10-09)
A new colorimetric sensor for sensing Hg2+ in a high-salinity solution has been developed using gold nanoparticles (AuNPs) decorated with 3-mercaptopropionate acid (MPA) and adenosine monophosphate (AMP). Because of the high negative charge density of AMP on each AuNP surface
Self-assembly of short and long-chain n-alkyl thiols onto gold surfaces: A real-time study using surface plasmon resonance techniques
Debono, Reno F., et al
Canadian Journal of Chemistry, 74, 677-688 (1996)
Vasileios Drogkaris et al.
ChemPlusChem, 85(11), 2466-2474 (2020-11-18)
The nucleophile-initiated mechanism of thiol-Michael reactions naturally leads to the formation of undesired nucleophile byproducts. Three aza-Michael compounds representing nucleophile byproducts of thiol-acrylate reactions initiated by 4-dimethylaminopyridine (DMAP), 1-methylimidazole (MIM), and 1,8-diazabicyclo[5.4.0]undec-7-ene (DBU) have been synthesized and their reactivity in
Tesfalem Geremariam Welearegay et al.
Nanotechnology, 29(6), 065603-065603 (2017-12-06)
Here, we report for the first time the fabrication of ligand-functionalised ultrapure monodispersed metal nanoparticles (Au, Cu, and Pt) from their pure metal precursors using the advanced gas deposition technique. The experimental conditions during nanoparticle formation were adjusted in order

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service