Skip to Content
Merck
All Photos(1)

Key Documents

40460

Sigma-Aldrich

N,N-Dimethylhexadecylamine

technical, ≥95% (GC)

Synonym(s):

1-(Dimethylamino)hexadecane, N,N-Dimethylpalmitylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)15N(CH3)2
CAS Number:
Molecular Weight:
269.51
Beilstein:
1755921
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

grade

technical

Quality Level

Assay

≥95% (GC)
≥98.0% (NT)

impurities

≤0.3% water

density

0.801 g/mL at 20 °C (lit.)

functional group

amine

SMILES string

CCCCCCCCCCCCCCCCN(C)C

InChI

1S/C18H39N/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19(2)3/h4-18H2,1-3H3

InChI key

NHLUVTZJQOJKCC-UHFFFAOYSA-N

General description

N,N-Dimethylhexadecylamine is a swelling agent useful for the enlargement of the size of mesopores of colloidal silica nanoparticles.[1]

Application

N,N-Dimethylhexadecylamine may be employed in the following studies:
  • As pore size mediator for the synthesis of monodispersed mesoporous silica nanoparticles.[2]
  • As swelling agent in the fabrication of colloidal mesoporous silica nanoparticles of a tunable mesopore size.[1]
  • Preparation of 1,4-bis(N-hexadecyl-N,N-dimethylammonium)butane dibromide, Gemini surfactant.[3]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

287.6 °F - closed cup

Flash Point(C)

142 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amani L Lee et al.
ACS nano, 11(6), 5623-5632 (2017-05-16)
Oxygen homeostasis is important in the regulation of biological function. Disease progression can be monitored by measuring oxygen levels, thus producing information for the design of therapeutic treatments. Noninvasive measurements of tissue oxygenation require the development of tools with minimal
Ramiro Martínez-Cámara et al.
Folia microbiologica, 65(3), 523-532 (2019-12-14)
Bacteria have developed different intra- and inter-specific communication mechanisms that involve the production, release, and detection of signaling molecules, because these molecules serve as the autoinducers involved in "quorum sensing" systems. Other communication mechanisms employ volatile signaling molecules that regulate
Ernesto Vázquez-Chimalhua et al.
Protoplasma, 256(3), 643-654 (2018-11-02)
Chemical communication underlies major adaptive traits in plants and shapes the root microbiome. An increasing number of diffusible and/or volatile organic compounds released by bacteria have been identified, which play phytostimulant or protective functions, including dimethyl-hexa-decylamine (DMHDA), a volatile biosynthesized
Shanshan Dai et al.
Physical chemistry chemical physics : PCCP, 20(16), 11285-11295 (2018-04-11)
A series of CO2-responsive oil-in-water (O/W) emulsions were prepared by introducing hydrophobic tertiary amines (TAs) with varying alkane carbon numbers (ACNs) into the emulsion stabilized by sodium dodecyl benzene sulfonate (SDBS). TAs are converted to bicarbonate salts upon bubbling of
Jinlou Gu et al.
Journal of colloid and interface science, 407, 236-242 (2013-07-20)
Despite their great potentials as biomacromolecues delivery vehicles, there are few, if any, reports on mesoporous silica nanoparticles (MSNs) simultaneously integrated with the merits of large pore size, small particle diameters and well-ordered mesostructure. Here, we designed a facile strategy

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service