Skip to Content
Merck
All Photos(2)

Documents

399191

Sigma-Aldrich

Rhodium(II) trifluoroacetate dimer

Synonym(s):

Trifluoroacetic acid rhodium(II) salt dimer

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CF3COO)2Rh]2
CAS Number:
Molecular Weight:
657.87
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

form

powder

reaction suitability

reagent type: catalyst

SMILES string

FC(F)(F)C(=O)O[Rh]OC(=O)C(F)(F)F.FC(F)(F)C(=O)O[Rh]OC(=O)C(F)(F)F

InChI

1S/4C2HF3O2.2Rh/c4*3-2(4,5)1(6)7;;/h4*(H,6,7);;/q;;;;2*+2/p-4

InChI key

PQEXTYUESSEHKW-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Application

Used in the preparation of isomerically pure α,β-unsaturated carbonyl compounds and the preparation of building blocks for one-, two-, and three-dimensional molecular solids.

Other Notes

Contains a trace amount of rhodium acetate

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li-Feng Mao et al.
Inorganic chemistry, 35(11), 3183-3187 (1996-05-22)
The complexes trans-[MI(2)(CNC(6)H(4)-CN-4)(2)], (M = Pd and Pt), trans-[FeI(2)L(4)] (L = CNC(6)H(4)-CN-4 and CNC(6)H(2)-Me(2)-2,6-CN-4), and [Mn(CNC(6)H(4)-CN-4)(6)][SO(3)CF(3)] were prepared. The compounds are thermally stable up to 230 degrees C or higher. The molecular structure of trans-[FeI(2)(CNC(6)H(4)-CN-4)(4)] was determined by X-ray crystallography:
A Convenient Method for the Preparation of (Z)-alpha,beta-Unsaturated Carbonyl Compounds.
Douglass F. Taber et al.
The Journal of organic chemistry, 61(8), 2908-2910 (1996-04-19)
Adam J Biacchi et al.
ACS nano, 9(2), 1707-1720 (2015-01-30)
The shapes of noble metal nanoparticles directly impact their properties and applications, including in catalysis and plasmonics, and it is therefore important to understand how multiple distinct morphologies can be controllably synthesized. Solution routes offer powerful capabilities for shape-controlled nanoparticle

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service