Skip to Content
Merck
All Photos(1)

Key Documents

395404

Sigma-Aldrich

Titanium(IV) chloride tetrahydrofuran complex

97%

Synonym(s):

Tetrachlorobis(tetrahydrofuran)titanium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TiCl4 · 2THF
CAS Number:
Molecular Weight:
333.89
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

97%

reaction suitability

core: titanium
reagent type: Lewis acid
reagent type: catalyst

mp

126-128 °C (lit.)

functional group

ether

storage temp.

2-8°C

SMILES string

Cl[Ti](Cl)(Cl)Cl.C1CCOC1.C2CCOC2

InChI

1S/2C4H8O.4ClH.Ti/c2*1-2-4-5-3-1;;;;;/h2*1-4H2;4*1H;/q;;;;;;+4/p-4

InChI key

LXWBMENBONGPSB-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

General description

Titanium(IV) chloride tetrahydrofuran complex, also known as TiCl4-THF, is a commonly used reagent in organic synthesis due to its Lewis acidic properties. It is used as a catalyst in various Friedel-Crafts reactions for the synthesis of aromatic compounds.

Application

Titanium(IV) chloride tetrahydrofuran complex can be used as a catalyst in intermolecular hydroamination reactions.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service