Skip to Content
Merck
All Photos(2)

Documents

393622

Sigma-Aldrich

p-Tolylboronic acid

97%

Synonym(s):

(p-Methylphenyl)boronic acid, 4-Methylbenzeneboronic acid, 4-Methylphenylboronic acid, 4-Tolueneboronic acid, 4-Tolylboronic acid, p-Tolueneboronic acid, NSC 62870, p-Methylbenzeneboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4B(OH)2
CAS Number:
Molecular Weight:
135.96
Beilstein:
2935970
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

mp

256-263 °C (lit.)

SMILES string

Cc1ccc(cc1)B(O)O

InChI

1S/C7H9BO2/c1-6-2-4-7(5-3-6)8(9)10/h2-5,9-10H,1H3

InChI key

BIWQNIMLAISTBV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reagent used for
  • Palladium (Pd)-catalyzed direct arylation
  • Direct Palladium(II)-Catalyzed Synthesis
  • Palladium-catalyzed arylation by Suzuki-Miyaura cross-coupling in water
  • Cyclopalladation
  • Tandem-type Pd(II)-catalyzed oxidative Heck reaction and intramolecular C-H amidation sequence
  • Ruthenium catalyzed direct arylation
  • Rhodium-catalyzed asymmetric conjugate addition
  • Ligand-free copper-catalyzed cross-coupling reactions
  • Regioselective arylation and alkynylation by Suzuki-Miyaura and Sonogashira cross-coupling reactions
  • Ligand-free Suzuki, Sonogashira, and Heck cross-coupling reactions

Reagent used in Preparation of
  • Catalysts for Suzuki-Miyaura cross-coupling of aryl bromides
  • Recyclable Palladium nanoparticle catalysts immobilized by click ionic copolymers as for Suzuki-Miyaura cross-coupling reactions in water

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

ACS Symp. Ser., 642, 194-205 (1996)
Jiajia Tan et al.
Biomaterials, 178, 608-619 (2018-04-02)
Inflammation serves as a natural defense mechanism to protect living organisms from infectious diseases. Nonsteroidal anti-inflammatory drugs (NSAIDs) can help relieve inflammatory reactions and are clinically used to treat pain, fever, and inflammation, whereas long-term use of NSAIDs may lead
Palladium nanoparticles immobilized by click ionic copolymers: Efficient and recyclable catalysts for Suzuki-Miyaura cross-coupling reaction in water
Zhang, D.; Zhou, C.; Wang, R.
Catalysis Communications, 22, 83-88 (2012)
Daniel P O'Connell et al.
Bioorganic & medicinal chemistry letters, 22(3), 1397-1401 (2012-01-10)
Endothelial lipase (EL) and lipoprotein lipase (LPL) are homologous lipases that act on plasma lipoproteins. EL is predominantly a phospholipase and appears to be a key regulator of plasma HDL-C. LPL is mainly a triglyceride lipase regulating (V)LDL levels. The
Carbon-Carbon Bond Formation by Ligand-free Cross-Coupling Reaction Using Palladium Catalyst Supported on Synthetic Adsorbent
Monguchi, Y.; et al.
ChemCatChem, 4, 546-558 (2012)

Articles

MIDA-protected boronate esters offer stability, chromatography compatibility, and reactivity in anhydrous cross-coupling conditions.

MIDA-protected boronate esters offer stability, chromatography compatibility, and reactivity in anhydrous cross-coupling conditions.

MIDA-protected boronate esters offer stability, chromatography compatibility, and reactivity in anhydrous cross-coupling conditions.

MIDA-protected boronate esters offer stability, chromatography compatibility, and reactivity in anhydrous cross-coupling conditions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service