Skip to Content
Merck
All Photos(3)

Documents

392219

Sigma-Aldrich

Cerium(III) nitrate hexahydrate

99.99% trace metals basis

Synonym(s):

Cerium trinitrate, Cerous nitrate hexahydrate, Nitric acid cerium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ce(NO3)3 · 6H2O
CAS Number:
Molecular Weight:
434.22
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.99% trace metals basis

form

crystals and lumps

reaction suitability

reagent type: catalyst
core: cerium

impurities

≤150.0 ppm Trace Rare Earth Analysis

SMILES string

[Ce+3].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/Ce.3NO3.6H2O/c;3*2-1(3)4;;;;;;/h;;;;6*1H2/q+3;3*-1;;;;;;

InChI key

QQZMWMKOWKGPQY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cerium (III) nitrate hexahydrate is a white-to-yellowcrystalline salt, which is hygroscopic and air-sensitive. Cerium (III) nitratehexahydrate is highly soluble in water, alcohol, and acetone. Cerium (III)nitrate hexahydrate melts at 57 °C and begins to thermally decompose at190 °Cwith complete decomposition to the oxide by 400 °C.

Application

Cerium (III) nitrate hexahydrate is widely used as a sourceof cerium, especially in the synthesis of micro- or nano-structured ceria(cerium oxide). Researchers leverage the high solubility and low decompositiontemperature of cerium (III) nitrate hexahydrate in hydrothermal reactions,sol-gel processing, and co-precipitation/calcination reactions to synthesizeceria and cerium-compounds. For example, for one study materials scientistsprepared a two-dimensional cobalt-doped ceria nanosheet by co-precipitating andcalcining cerium nitrate and cobalt acetate. As another example, researchersused the high solubility of cerium nitrate to grow crystals of two-dimensionalrare-earth double perovskites. Researchers have also used cerium nitrate to formcomposites, such as graphene-ceria nanoparticle composites and TiO2/CeO2nanocomposites, utilizing cerium’s (III/IV) redox chemistry forlight-absorption and photocatalysis. One example, that shows the maturematerials engineering of cerium (III) nitrate chemistry, is a study that uses acalcium-doped cerium nanotube composite with lithiumbis(trifluoromethanesulfonyl)imide (LiTFSI) poly(ethylene oxide) for asolid-state lithium-ion electrolyte. The Ca-CeO2 nanotubes, which achievehigher concentrations of oxygen vacancies than undoped CeO2, were made byelectrospinning from a solution of cerium nitrate and calcium nitrate withpolyvinylpyrrolidone, which is removed by pyrolysis in a final calcination.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shan Gao et al.
Small (Weinheim an der Bergstrasse, Germany), 16(19), e1906668-e1906668 (2020-04-17)
A high efficiency and great tunability of bandwidth and absorption-range electromagnetic wave absorber is proposed without precedent. A series of 2D carbon-based nanocomposites with the loading of cerium oxide (CN-Ce) and other types of rare earth oxides (CN-REOs) can be
Chao Shi et al.
Journal of the American Chemical Society, 142(1), 545-551 (2019-12-12)
As a major branch of hybrid perovskites, two-dimensional (2D) hybrid double perovskites are expected to be ideal systems for exploring novel ferroelectric properties, because they can accommodate a variety of organic cations and allow diverse combinations of different metal elements.
Shuaihu Jiang et al.
Journal of the American Chemical Society, 142(14), 6461-6466 (2020-03-24)
As an alternative for depleting fossil fuel energy, hydrogen economy desires low-cost and efficient hydrogen production from water splitting. In order to explore a cheap, abundant, active, and durable catalyst for the electrocatalytic hydrogen evolution reaction (HER), two-dimensional (2D) ceria
Stable Seamless Interfaces and Rapid Ionic Conductivity of Ca?CeO2/LiTFSI/PEO Composite Electrolyte for High-Rate and High-Voltage All-Solid-State Battery.
Chen H, et al.
Advanced Energy Materials, 10, 2000049-2000049 (2020)
Synergetic Effect between Photocatalysis on TiO2 and Thermocatalysis on CeO2 for Gas-Phase Oxidation of Benzene on TiO2/CeO2 Nanocomposites.
Zeng M, et al.
ACS Catalysis, 5 (6), 3278-3286 (2015)

Articles

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service