Skip to Content
Merck
All Photos(2)

Key Documents

389056

Sigma-Aldrich

3,5-Dichlorophenyl isocyanate

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3NCO
CAS Number:
Molecular Weight:
188.01
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

form

solid

mp

32-34 °C (lit.)

density

1.38 g/mL at 25 °C (lit.)

SMILES string

Clc1cc(Cl)cc(c1)N=C=O

InChI

1S/C7H3Cl2NO/c8-5-1-6(9)3-7(2-5)10-4-11/h1-3H

InChI key

XEFUJGURFLOFAN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3,5-Dichlorophenyl isocyanate can be used:
  • As a reactant to synthesize 3-(3,5-Dichlorophenyl)-2,4-oxazolidinedione (DCPO).
  • To synthesize phenylcarbamate derivatives of cellulose and amylose, which are useful as polysaccharide-based chiral stationary phases (CSPs).{70]

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Aquatic Chronic 2 - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of regioselectively substituted cellulose derivatives and applications in chiral chromatography.
Acemoglu M, et al.
Chirality, 10(4), 294-306 (1998)
Nephrotoxic and hepatotoxic potential of imidazolidinedione-, oxazolidinedione-and thiazolidinedione-containing analogues of N-(3, 5-dichlorophenyl) succinimide (NDPS) in Fischer 344 rats.
Kennedy E, et al.
Toxicology, 186(1-2), 79-91 (2003)
Synthesis and chiral recognition of amylose derivatives bearing regioselective phenylcarbamate substituents at 2, 6-and 3-positions for high-performance liquid chromatography.
Shen J, et al.
Journal of Chromatography A, 1467, 199-205 (2016)
Polymer?bound cellulose phenylcarbamate derivatives as chiral stationary phases for enantioselective HPLC.
Ling F, et al.
Journal of Separation Science, 26(15?16), 1337-1346 (2003)
Alyssa M Keil et al.
Toxicology in vitro : an international journal published in association with BIBRA, 29(7), 1887-1896 (2015-07-21)
Liver damage occurred in some patients who took troglitazone (TGZ) for type II diabetes. The 2,4-thiazolidinedione (TZD) ring in TGZ's structure has been implicated in its hepatotoxicity. To further examine the potential role of a TZD ring in toxicity we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service