Skip to Content
Merck
All Photos(1)

Key Documents

374857

Sigma-Aldrich

3,5-Bis(trifluoromethyl)phenyl isocyanate

98%

Synonym(s):

1-Isocyanato-3,5-bis(trifluoromethyl)benzene, 3,5-Di(trifluoromethyl)phenyl isocyanate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF3)2C6H3NCO
CAS Number:
Molecular Weight:
255.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.43 (lit.)

density

1.476 g/mL at 25 °C (lit.)

functional group

fluoro
isocyanate

SMILES string

FC(F)(F)c1cc(cc(c1)C(F)(F)F)N=C=O

InChI

1S/C9H3F6NO/c10-8(11,12)5-1-6(9(13,14)15)3-7(2-5)16-4-17/h1-3H

InChI key

NRSSOFNMWSJECS-UHFFFAOYSA-N

Application

3,5-Bis(trifluoromethyl)phenyl isocyanate may be used in following studies:
  • Chemical derivatization of amino-functionalized model surfaces.
  • Preparation of arylaminothiocarbonylpyridinium zwitterionic salts, via an exothermic reaction with 4-pyrrolidinopyridine.
  • Synthesis of (S)-3-hydroxymethyl-2-methoxymethoxy-2′-(3-(3,5-bis(trifluoromethyl)phenyl)uryl-benzyl)-1,10-binaphthalene.
  • Synthesis of 1-[3,5-bis(trifluoromethyl) phenyl]-3-(2-pyridyl)thiourea, via reaction with 2-amino pyridine in MeCN.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

154.4 °F - closed cup

Flash Point(C)

68 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kazuaki Ishihara et al.
Organic letters, 10(11), 2187-2190 (2008-04-30)
The exothermic reaction of 3,5-bis(trifluoromethyl)phenyl or 4-nitrophenyl isothiocyanate with 4-pyrrolidinopyridine (PPY) gave the corresponding arylaminothiocarbonylpyridinium salts in quantitative yields. These novel zwitterionic salts were effective as organocatalysts for the transesterification reaction of an equimolar mixture of methyl carboxylates and alcohols
Huadong Yue et al.
Acta crystallographica. Section E, Structure reports online, 64(Pt 5), o858-o858 (2008-01-01)
The title compound, C(14)H(9)F(6)N(3)S, exhibits a nearly planar conformation in the solid state, with a dihedral angle between the planes of the benzene and pyridine rings of 14.86 (3)°. The pyridine N atom allows for the formation of a six-membered N-H⋯N(py)
Utsab Manna et al.
Dalton transactions (Cambridge, England : 2003), 46(35), 11956-11969 (2017-08-31)
A rationally designed ortho-phenylenediamine based trifluoromethyl meta-disubstituted bis-urea receptor (L) exhibits effective, consistent and systematic binding in its neutral form towards smaller spherical halides (fluoride, chloride, bromide and iodide), and relatively larger planar carbonate and tetrahedral sulphate oxyanions. All the
Effects of ring substituents on enantioselective recognition of amino alcohols and acids in uryl-based binol receptors.
Nandhakumar R, et al.
Tetrahedron, 64(33), 7704-7708 (2008)
Biao Wang et al.
The Science of the total environment, 710, 135512-135512 (2019-12-02)
The excessive use of acetochlor (ACT), a commonly used herbicide with latent endocrine disrupting functions, causes surface water pollution. The efficient removal of ACT from contaminated water supplies is of paramount importance. In the current work, 3,5-Bis(trifluoromethyl)phenyl isocyanate (BTP) was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service