Skip to Content
Merck
All Photos(1)

Key Documents

388548

Sigma-Aldrich

Strontium acetate

Synonym(s):

Strontium diacetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CO2)2Sr
CAS Number:
Molecular Weight:
205.71
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22

impurities

~3% water

SMILES string

CC(=O)O[Sr]OC(C)=O

InChI

1S/2C2H4O2.Sr/c2*1-2(3)4;/h2*1H3,(H,3,4);/q;;+2/p-2

InChI key

RXSHXLOMRZJCLB-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Strontium acetate can be used as a strontium precursor to synthesize:
  • Strontium titanate (SrTiO3) via hydrothermal synthesis with titanium isopropoxide.
  • Lead strontium titanate thin films, applicable for the development of some tunable dielectric devices.
  • Strontium cyanamide polymorphs via ammonia nitridation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A facile hydrothermal synthesis of SrTiO3 for dye sensitized solar cell application
Jayabal P, et al.
Journal of alloys and compounds, 586, 456-461 (2014)
Ammonia nitridation synthesis and structural change of strontium cyanamide polymorphs
Takeda T, et al.
J. Ceram. Soc. Jpn., 115(1347), 729-731 (2007)
Charles R Parkinson et al.
The Journal of clinical dentistry, 22(3), 74-81 (2011-09-13)
The objective of this study was to evaluate the relative level of dentin tubule occlusion and dentin mineralization conferred by a 5% w/w calcium sodium phosphosilicate (45S5)/1450 ppm fluoride toothpaste in comparison to a range of commercial toothpastes reported to
R Patel et al.
The Journal of clinical dentistry, 22(4), 108-112 (2011-01-01)
The in vitro effects of two commercial sensitivity relief dentifrices, one containing 8.0% arginine, calcium carbonate, and 1450 ppm fluoride as sodium monofluorophosphate (MFP), and the other containing 8% strontium acetate and 1040 ppm fluoride as sodium fluoride, in occluding
T Schiff et al.
The Journal of clinical dentistry, 22(4), 128-138 (2011-01-01)
The objective of this 16-week, double-blind, randomized, switch-over design study was to compare the efficacy in reducing dentin hypersensitivity of a dentifrice containing 8.0% arginine, calcium carbonate, and 1450 ppm fluoride as sodium monofluorophosphate (Colgate Sensitive Pro-Relief [also marketed as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service