Skip to Content
Merck
All Photos(4)

Key Documents

377635

Sigma-Aldrich

Octadecyl-functionalized silica gel

200-400 mesh, extent of labeling: 20-22% loading

Synonym(s):

C18 silica gel

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
NACRES:
NA.22

form

powder

extent of labeling

20-22% loading

surface area

~550 m2/g

matrix

irregular silica particle platform

matrix active group

C18 phase

particle size

200-400 mesh

pore size

60 Å pore size

Looking for similar products? Visit Product Comparison Guide

Application

Octadecyl-functionalized silica gel is generally used as a stationary phase for low- and medium-pressure reversed-phase liquid chromatography.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and characterization of polymeric C18 stationary phases for liquid chromatography
Sander LC and Wise SA
Analytical Chemistry, 56(3), 504-510 (1984)
Antitrypanosomatid activity of flavonoid glycosides isolated from Delphinium gracile, D. staphisagria, Consolida oliveriana and from Aconitum napellus subsp. Lusitanicum
Marin C, et al.
Phytochemistry Letters, 19(2), 196-209 (2017)
Acylated flavonol glycosides from Delphinium staphisagria
Diaz JG, et al.
Phytochemistry Letters, 1(2), 125-129 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service