Skip to Content
Merck
All Photos(1)

Key Documents

377481

Sigma-Aldrich

Isopropyltriphenylphosphonium iodide

98%

Synonym(s):

(1-Methylethyl)triphenylphosphonium iodide, NSC 140615

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHP(C6H5)3I
CAS Number:
Molecular Weight:
432.28
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

reaction suitability

reaction type: C-C Bond Formation

mp

194-197 °C (lit.)

SMILES string

[I-].CC(C)[P+](c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/C21H22P.HI/c1-18(2)22(19-12-6-3-7-13-19,20-14-8-4-9-15-20)21-16-10-5-11-17-21;/h3-18H,1-2H3;1H/q+1;/p-1

InChI key

HHBXWXJLQYJJBW-UHFFFAOYSA-M

Application

Employed in Wittig and cyclopropanation reactions.
Reactant for:
  • Total synthesis of heliananes
  • Preparation of highly substituted benzene derivatives
  • Synthesis of curcuphenol and elvirol analogs via a retro-aldol reaction as fungicidal agents
  • 4-substituted methoxylbenzoyl-aryl-thiazoles analogues as potent and orally bioavailable anticancer agents
  • Wittig reaction

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of the Chemical Society. Perkin Transactions 1, 3211-3211 (1992)
Tetrahedron Letters, 34, 2695-2695 (1993)
Journal of the Chemical Society. Perkin Transactions 1, 31-31 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service