Skip to Content
Merck
All Photos(2)

Key Documents

372358

Sigma-Aldrich

Triacetoxy(methyl)silane

technical grade, 90%

Synonym(s):

Methyltriacetoxysilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CO2)3SiCH3
CAS Number:
Molecular Weight:
220.25
Beilstein:
1788668
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

90%

form

solid

bp

94-95 °C/9 mmHg (lit.)

mp

40-45 °C (lit.)

density

1.20 g/mL at 20 °C (lit.)

SMILES string

CC(=O)O[Si](C)(OC(C)=O)OC(C)=O

InChI

1S/C7H12O6Si/c1-5(8)11-14(4,12-6(2)9)13-7(3)10/h1-4H3

InChI key

TVJPBVNWVPUZBM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

183.2 °F - closed cup

Flash Point(C)

84 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Udagama et al.
The Journal of prosthetic dentistry, 48(1), 86-88 (1982-07-01)
The urethanes, PVC, acrylic resins, and silicones are still the most popular among a large number of materials being used to fabricate facial prostheses. Each material has advantages and disadvantages. The ideal material is not yet available. It was found
Silicone Vapor Poisoning of Catalytic Methane Sensors
Chilton, J. E., et al.
Proceedings of the fourth WVU Conference on Coal Mine Electrotechnology (1978)
Solvation of lithium salts within single-phase dimethyl siloxane bisphenol-A carbonate block copolymer.
Spiegel EF, et al.
Polymer, 41(9), 3365-3369 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service