Skip to Content
Merck
All Photos(3)

Key Documents

367753

Sigma-Aldrich

Cerium(III) acetate hydrate

99.9% trace metals basis

Synonym(s):

Cerium acetate hydrate, Cerium triacetate, Cerium(3+) triacetate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ce(CH3CO2)3 · xH2O
CAS Number:
Molecular Weight:
317.25 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Quality Level

Assay

99.9% trace metals basis

form

powder and chunks

composition

Degree of hydration, 1-3

reaction suitability

core: cerium
reagent type: catalyst

impurities

≤1% acetic acid

SMILES string

O.CC(=O)O[Ce](OC(C)=O)OC(C)=O

InChI

1S/3C2H4O2.Ce.H2O/c3*1-2(3)4;;/h3*1H3,(H,3,4);;1H2/q;;;+3;/p-3

InChI key

AERUOEZHIAYQQL-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

With bromide ion, catalyzes the liquid-phase auto-oxidation of cresols.[1][2]Cerium (III) acetate hydrate may be used as the starting material for the synthesis of CeO2 nanoparticles.[3]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chem. Abstr., 112, 219165x-219165x (1990)
Thermal decomposition of cerium (III) acetate studied with sample-controlled thermogravimetric?mass spectrometry (SCTG?MS).
Arii T,et al.
J. Eur. Ceram. Soc., 22(13), 2283-2289 (2002)
Synthesis, structural and optical properties of CeO 2 nanoparticles synthesized by a simple polyvinyl pyrrolidone (PVP) solution route.
Phoka S, et al.
Materials Chemistry and Physics, 115(1), 423-428 (2009)
Nippon Kagaku Kaishi, 92-92 (1990)

Articles

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service