Skip to Content
Merck
All Photos(3)

Key Documents

363340

Sigma-Aldrich

N-Acetylcysteamine

95%

Synonym(s):

N-(2-Mercaptoethyl)acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CONHCH2CH2SH
CAS Number:
Molecular Weight:
119.19
MDL number:
UNSPSC Code:
12352100
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

liquid

refractive index

n20/D 1.511 (lit.)

bp

138-140 °C/7 mmHg (lit.)

mp

6-7 °C (lit.)

density

1.121 g/mL at 25 °C (lit.)

functional group

amide
thiol

SMILES string

CC(=O)NCCS

InChI

1S/C4H9NOS/c1-4(6)5-2-3-7/h7H,2-3H2,1H3,(H,5,6)

InChI key

AXFZADXWLMXITO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N-Acetylcysteamine, also known as N-(2-Mercaptoethyl) acetamide, is a derivative of cysteamine, that is commonly used as a building block for the synthesis of alkylated thiol and thioesters via esterification.

Application

N-Acetylcysteamine can be used as a building block to synthesize:
  • N







  • -acetylcysteamine (SNAC) thioesters by reacting with various acid derivatives in the presence of 1,1′-carbonyldiimidazole (CDI).      
  • Thieno[2,3-c]pyrrole derivatives via three-component reaction of 2-acetyl-3-thiophenecarboxaldehyde and various amines.
  •  Carbapenems, a class of beta-lactam antibiotic agents.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isolation, properties, and regulation of a mitochondrial acyl coenzyme A thioesterase from pig heart.
K Y Lee et al.
The Journal of biological chemistry, 254(11), 4516-4523 (1979-06-10)
N Singh et al.
Biochemical and biophysical research communications, 131(2), 786-792 (1985-09-16)
The acetyl transacylase activity of the fatty acid synthase from yeast has been investigated using p-nitrophenylthiol acetate. The chromophoric nature of the nitrophenylthiol moiety affords a convenient spectrophotometric assay for the transacylase function as well as a means to investigate
Magoichi Sako et al.
The Journal of organic chemistry, 63(20), 6947-6951 (2001-10-24)
4H-[1,2,5]Oxadiazolo[3,4-d]pyrimidine-5,7-dione 1-oxides (2) are conveniently prepared in high yields by the oxidative intramolecular cyclization of 6-amino-5-nitro-1H-pyrimidine-2,4-diones (1) employing iodosylbenzene diacetate as an oxidant in the presence of lithium hydride. The generation of nitric oxide (NO) and NO-related species from 2
Micah J Bodner et al.
Organic letters, 11(16), 3606-3609 (2009-07-21)
Efficient syntheses of N-acetyl thienamycin and epithienamycin A in their readily deprotected form are reported where three contiguous stereocenters are established in a single catalytic asymmetric azetidinone-forming reaction. These examples are a template for synthesizing C-5/C-6 cis or trans carbapenems
Cell cycle arrest and apoptosis induction by an anticancer chalcone epoxide
Haiyong H, et al
Archives of Pharmacal Research, 343, 429-439 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service