Skip to Content
Merck
All Photos(1)

Key Documents

359734

Sigma-Aldrich

Calcium citrate tetrahydrate

99%

Synonym(s):

1,2,3-Propanetricarboxylic acid, 2-hydroxy-, calcium salt (2:3), tetrahydrate, Tricalcium dicitrate tetrahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[O2CCH2C(OH)(CO2)CH2CO2]2Ca3 · 4H2O
CAS Number:
Molecular Weight:
570.49
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

powder

SMILES string

[H]O[H].[H]O[H].[H]O[H].[H]O[H].OC1(CC(=O)O[Ca]OC(=O)C1)C(=O)O[Ca]OC(=O)C2(O)CC(=O)O[Ca]OC(=O)C2

InChI

1S/2C6H8O7.3Ca.4H2O/c2*7-3(8)1-6(13,5(11)12)2-4(9)10;;;;;;;/h2*13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);;;;4*1H2/q;;3*+2;;;;/p-6

InChI key

LNIZKKFWMDARJV-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Thermal decomposition of calcium citrate tetrahydrate in dynamic air or dry nitrogen has been investigated by by TG, DTA and DSC.

Application

Calcium citrate tetrahydrate may be used as raw material in the preparation of nanoparticle with the nominal composition Ca0.8 Ba0.2 Ti03 :Pr3+, via sol-gel process. It may be employed as carbon source in a simple and effective carbonization method to prepare porous carbon without any activation process. It is suitable for use in the preparation of bovine serum albumin (BSA) nanoparticles.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yannan Mu et al.
Luminescence : the journal of biological and chemical luminescence, 30(5), 503-506 (2014-09-12)
Nanoparticles with the nominal composition Ca0.8Ba0.2Ti03:Pr(3+) were prepared using the sol-gel process. Barium nitrate, 4-hydrated calcium nitrate and praseodymium oxide were used as raw materials. The structural evolution and decomposition processes of the precursors were investigated by powder X-ray diffraction
Thermal decomposition of calcium citrate tetrahydrate.
Mansour SAA.
Thermochimica Acta, 233(2), 243-256 (1994)
Preparation of size-controlled bovine serum albumin (BSA) nanoparticles by a modified desolvation method.
Jun JY, et al.
Food Chemistry, 127(4), 1892-1898 (2011)
An activation-free protocol for preparing porous carbon from calcium citrate and the capacitive performance.
Zhou QQ, et al.
Microporous and Mesoporous Materials : The Official Journal of the International Zeolite Association, 158, 155-161 (2012)
Barbara Dolinska et al.
Biological trace element research, 142(3), 456-464 (2010-09-03)
An in vitro model was used to simulate the intestinal permeation of calcium ions depending on the type of salt (carbonate, fumarate, citrate, or gluconate), its concentration (1.0, 2.5, 5.0, or 10 mM/l), and pH (1.3, 4.2, 6.2, or 7.5). To

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service