Skip to Content
Merck
All Photos(1)

Documents

359297

Sigma-Aldrich

Sodium ethanethiolate

technical grade, 80%

Synonym(s):

Ethanethiol sodium salt, Ethyl mercaptan sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2SNa
CAS Number:
Molecular Weight:
84.12
Beilstein:
3593647
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Quality Level

Assay

80%

SMILES string

CCS[Na]

InChI

1S/C2H6S.Na/c1-2-3;/h3H,2H2,1H3;/q;+1/p-1

InChI key

QJDUDPQVDAASMV-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Reaction of FeCl2 or FeCl3 with sodium ethanethiolate in N-methylformamide (NMF) has been evaluated. Reaction of sodium ethanethiolate with substituted 1,2-dithiole-3-thiones has been studied.

Application

Sodium ethanethiolate may be used in the equilibrium titrations in N-methylformamide of G-25 gel filtered (ox)-state FeMo cofactor [FeMoco(ox)] from Azotobacter vinelandii nitrogenase. It may be used in the synthesis of a series of hybrid compounds combining the pharmacophoric redox moieties of vitamin E and key features responsible for the antiarrhythmic properties of the class I antiarrhythmics procainamide and lidocaine.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Frank et al.
Journal of biological inorganic chemistry : JBIC : a publication of the Society of Biological Inorganic Chemistry, 6(7), 683-697 (2001-10-30)
Equilibrium titrations in N-methylformamide (NMF) of G-25 gel filtered (ox)-state FeMo cofactor [FeMoco(ox)] from Azotobacter vinelandii nitrogenase were carried out using sodium ethanethiolate and followed using UV/Vis absorption spectroscopy. For Fe-Moco(ox), a non-linear least squares (NLLSQ) fit to the data
Patrick Frank et al.
Journal of biological inorganic chemistry : JBIC : a publication of the Society of Biological Inorganic Chemistry, 10(4), 373-382 (2005-05-03)
The reaction of FeCl(2) or FeCl(3) with sodium ethanethiolate (SEt) in N-methylformamide (NMF) has been reevaluated to rectify a previous Fe(II) oxidation artifact. On titrating Fe(II) with EtS(-) concentrations up to 12 mol Eq, new features in the UV/vis spectrum
Maria Koufaki et al.
Bioorganic & medicinal chemistry, 11(23), 5209-5219 (2003-11-08)
We have synthesized a series of hybrid compounds combining the pharmacophoric redox moieties of vitamin E and key features responsible for the antiarrhythmic properties of the class I antiarrhythmics procainamide and lidocaine. Procainamide analogue (2a) and lidocaine analogues (14a) and
Reactivity of substituted 1, 2-dithiole-3-thiones with sodium ethanethiolate: a convenient route to a novel heterocycle.
Largeron M, et al.
Tetrahedron, 43(15), 3421-3428 (1987)
Nariyasu Mano et al.
Analytical chemistry, 81(22), 9395-9401 (2009-10-23)
The analysis of post-translational phosphorylation is a crucial step in understanding the mechanisms of many physiological events. Numerous approaches for the development of analytical methods aimed at the detection and quantification of phosphorylated proteins by mass spectrometry have been reported

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service