Skip to Content
Merck
All Photos(1)

Documents

W425800

Sigma-Aldrich

Ethanethiol

≥97%

Synonym(s):

Ethyl mercaptan, Mercaptan C2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5SH
CAS Number:
Molecular Weight:
62.13
FEMA Number:
4258
Beilstein:
773638
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
12.017
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

Halal

reg. compliance

FDA 21 CFR 117

vapor density

2.1 (vs air)

vapor pressure

8.51 psi ( 20 °C)

Assay

≥97%

autoignition temp.

570 °F

expl. lim.

18.2 %

refractive index

n20/D 1.4306 (lit.)

bp

35 °C (lit.)

density

0.839 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fruity; sulfurous

storage temp.

2-8°C

SMILES string

CCS

InChI

1S/C2H6S/c1-2-3/h3H,2H2,1H3

InChI key

DNJIEGIFACGWOD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Liq. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-49.0 °F - closed cup

Flash Point(C)

-45 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Hanczkó et al.
Journal of chromatography. A, 1163(1-2), 25-42 (2007-07-04)
The main aims of this work were (a) to present the characteristics and stability of the o-phthalaldehyde (OPA)-ethanethiol (ET) derivatives of 22 amino acids, including the believed-to-be less stable OPA derivatives providing glycine, gamma-aminobutyric acid, beta-alanine, histidine, ornithine, lysine and
Edmanuel Torres et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 12(5), 999-1009 (2011-03-12)
The translational and orientational potential energy surfaces (PESs) of n-alkanethiols with up to four carbon atoms are studied for (√(3)×√(3))R30° self-assembled monolayers (SAMs). The PESs indicate that methanethiol may form SAM structures that are not accessible for long-chain thiols. The
A supramolecular network as sacrificial mask for the generation of a nanopatterned binary self-assembled monolayer.
Christophe Silien et al.
Small (Weinheim an der Bergstrasse, Germany), 6(3), 391-394 (2009-12-19)
Natalie J Galant et al.
The journal of physical chemistry. A, 113(32), 9138-9149 (2009-09-02)
First principle quantum molecular computations have been carried out at the B3LYP/6-31G(d,p) and G3MP2B3 levels of theory on ethyl mercaptan and diethyl disulfide to study their full conformational space. The consequences of molecular axis chirality for the potential energy hypersurface
Nariyasu Mano et al.
Analytical chemistry, 81(22), 9395-9401 (2009-10-23)
The analysis of post-translational phosphorylation is a crucial step in understanding the mechanisms of many physiological events. Numerous approaches for the development of analytical methods aimed at the detection and quantification of phosphorylated proteins by mass spectrometry have been reported

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service