All Photos(1)
About This Item
Linear Formula:
(CH3)3CCH(C6H5)OH
CAS Number:
Molecular Weight:
164.24
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
99%
bp
90 °C/5 mmHg (lit.)
mp
43-45 °C (lit.)
functional group
hydroxyl
phenyl
SMILES string
CC(C)(C)C(O)c1ccccc1
InChI
1S/C11H16O/c1-11(2,3)10(12)9-7-5-4-6-8-9/h4-8,10,12H,1-3H3
InChI key
YBVRFTBNIZWMSK-UHFFFAOYSA-N
General description
Raney nickel and Raney cobalt catalyzed transfer hydrogenolysis of 2,2-dimethyl-1-phenyl-1-propanol has been reported. Kinetic resolution of 2,2-dimethyl-1-phenyl-1-propanol using a lead dioxide anode modified with poly-S-valine grafted on to a polypyrrole film has been reported. An efficient Cu(I)-catalyzed oxidation of 2,2-dimethyl-1-phenyl-1-propanol with di-tert-butyldiaziridinone as oxidant under mild conditions has been investigated.
Application
2,2-Dimethyl-1-phenyl-1-propanol was used in the preparation of 2,2-dimethylpropiophenone.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
206.6 °F - closed cup
Flash Point(C)
97.00 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Poly (pyrroles) containing chiral side chains: effect of substituents on the chiral recognition in the doped as well as in the undoped state of the polymer film.
Pleus S and Schulte B.
Journal of Solid State Electrochemistry, 5(7-8), 522-520 (2001)
Brant Landers et al.
The Journal of organic chemistry, 76(5), 1390-1397 (2011-01-22)
The use of commercially available (SIPr)Pd(cinnamyl)Cl (SIPr = 1,3-bis(2,6-diisopropylphenyl)-4,5-dihydroimidazol-2-ylidene) as a precatalyst for the anaerobic oxidation of secondary alcohols is described. The use of this complex allows for a drastic reduction in the reaction times and catalyst loading when compared
Transfer hydrogenolysis of aromatic alcohols using Raney catalysts and 2-propanol.
Gross BH, et al.
Applied Catalysis A: General, 219(1), 281-289 (2001)
Yingguang Zhu et al.
Organic letters, 15(5), 992-995 (2013-02-19)
A novel and efficient Cu(I)-catalyzed oxidation of alcohols has been achieved with di-tert-butyldiaziridinone as the oxidant under mild conditions. A wide variety of primary and secondary alcohols with various functional groups can be oxidized to aldehydes and ketones in high
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service