Skip to Content
Merck
All Photos(2)

Key Documents

130362

Sigma-Aldrich

Isobutyrophenone

97%

Synonym(s):

2-Methyl-1-phenyl-1-propanone, Isobutyrylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COCH(CH3)2
CAS Number:
Molecular Weight:
148.20
Beilstein:
774499
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

1 mmHg ( 50 °C)

Assay

97%

form

liquid

refractive index

n20/D 1.517 (lit.)

bp

217 °C (lit.)

density

0.988 g/mL at 25 °C (lit.)

functional group

ketone
phenyl

SMILES string

CC(C)C(=O)c1ccccc1

InChI

1S/C10H12O/c1-8(2)10(11)9-6-4-3-5-7-9/h3-8H,1-2H3

InChI key

BSMGLVDZZMBWQB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Isobutyrophenone was used as an internal standard in the determination of hydroxyzine hydrochloride and benzyl alcohol in injection solutions. It was also used in the preparation of alpha-hydroxyisobutyrophenone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

183.2 °F

Flash Point(C)

84 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The chlorination of isobutyrophenone: determination of its p K a value and of the course of the reaction.
Guthrie JP and Cossar J.
Canadian Journal of Chemistry, 68(3), 397-403 (1990)
Zijun Wang et al.
Science advances, 6(39) (2020-09-27)
As a promising actuating material, liquid crystal elastomer (LCE) has been intensively explored in building diverse active structures and devices. Recently, direct ink writing technique has been developed to print LCE structures with various geometries and actuation behaviors. Despite the
Gavin McLaughlin et al.
Drug testing and analysis, 10(9), 1404-1416 (2018-04-20)
The availability of new psychoactive substances (NPS) on the recreational drug market continues to create challenges for scientists in the forensic, clinical and toxicology fields. Phenmetrazine (3-methyl-2-phenylmorpholine) and an array of its analogs form a class of psychostimulants that are
Won-Gyu Bae et al.
Science translational medicine, 11(503) (2019-08-02)
A flexible microneedle patch that can transdermally deliver liquid-phase therapeutics would enable direct use of existing, approved drugs and vaccines, which are mostly in liquid form, without the need for additional drug solidification, efficacy verification, and subsequent approval. Specialized dissolving
H Montazerian et al.
Acta biomaterialia, 96, 149-160 (2019-06-30)
In the present study, polydimethylsiloxane (PDMS) porous scaffolds are designed based on minimal surface architectures and fabricated through a low-cost and accessible sacrificial mold printing approach using a fused deposition modeling (FDM) 3D printer. The effects of pore characteristics on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service