Skip to Content
Merck
All Photos(1)

Documents

328057

Sigma-Aldrich

4,4′-Dibromobenzil

90%, technical grade

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4COCOC6H4Br
CAS Number:
Molecular Weight:
368.02
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Quality Level

Assay

90%

mp

224-226 °C (lit.)

SMILES string

Brc1ccc(cc1)C(=O)C(=O)c2ccc(Br)cc2

InChI

1S/C14H8Br2O2/c15-11-5-1-9(2-6-11)13(17)14(18)10-3-7-12(16)8-4-10/h1-8H

InChI key

NYCBYBDDECLFPE-UHFFFAOYSA-N

Gene Information

Related Categories

Application

4,4′-Dibromobenzil was used in preparation of 5,5′-bis -(4-bromo-phenyl)-3-methyl-imidazolidine-2,4-dione, 5,5′- bis -(4-bromo-phenyl)-3-butyl-imidazolidine-2,4-dione, 5,5′-bis-(4-bromo-phenyl)-3-pentyl-imidazolidine-2,4-dione and 5,5′-bis -(4-chloro-phenyl)-3-ethyl imidazolidine-2,4-dione. It was also used in synthesis of novel quinoxaline-based sensitizers for dye-sensitized solar cells.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dong Wook Chang et al.
Organic letters, 13(15), 3880-3883 (2011-06-28)
Novel quinoxaline-based organic sensitizers using vertical (RC-21) and horizontal (RC-22) conjugation between an electron-donating triphenylamine unit and electron-accepting quinoxaline unit have been synthesized and used for dye-sensitized solar cells (DSSCs), leading to the relatively high power conversion efficiencies of 3.30
A rapid and efficient microwave-assisted synthesis of hydantoins and thiohydantoins.
Muccioli GG, et al.
Tetrahedron, 59(8), 1301-1307 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service