Skip to Content
Merck
All Photos(2)

Documents

317837

Sigma-Aldrich

Dicyclohexylamine nitrite

97%

Synonym(s):

Dicyclohexylammonium nitrite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H11)2NH · HNO2
CAS Number:
Molecular Weight:
228.33
Beilstein:
3711911
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

3.3 mmHg ( 25 °C)

Quality Level

Assay

97%

form

powder

mp

182-183 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

ON=O.C1CCC(CC1)NC2CCCCC2

InChI

1S/C12H23N.HNO2/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12;2-1-3/h11-13H,1-10H2;(H,2,3)

InChI key

ZFAKTZXUUNBLEB-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Flam. Sol. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B G Feuerstein et al.
Cancer research, 45(10), 4950-4954 (1985-10-01)
Growth characteristics, polyamine levels, and distribution of cells in the cell cycle were determined for 9L rat brain tumor cells treated for various periods with 1 mM dicyclohexylamine sulfate (DCHA). Continuous treatment of cells with DCHA caused growth inhibition at
J L Mitchell et al.
Biochimica et biophysica acta, 840(3), 309-316 (1985-07-05)
Dicyclohexylamine, a spermidine synthase inhibitor, was evaluated for its ability to alter specific polyamine levels in rat hepatoma HTC cells in culture. Media concentrations of 0.5 and 1.0 mM reduced the production of spermidine from putrescine and enhanced the conversion
Massimo Donà et al.
Cancer research, 64(17), 6225-6232 (2004-09-03)
Hyperforin (Hyp), the major lipophilic constituent of St. John's wort, was assayed as a stable dicyclohexylammonium salt (Hyp-DCHA) for cytotoxicity and inhibition of matrix proteinases, tumor invasion, and metastasis. Hyp-DCHA triggered apoptosis-associated cytotoxic effect in both murine (C-26, B16-LU8, and
S J Pietr et al.
Mycoses, 32(7), 359-361 (1989-07-01)
Inhibitory doses (ID50) of phenols complexed with dicyclohexylamine are lower (21.4-47.7%) than the inhibitory doses of free phenols. This is demonstrated in vitro by experiments with Aspergillus oryzae and Penicillium expansum on solid medium.
A F Tiburcio et al.
Plant cell, tissue and organ culture, 9, 111-120 (1987-01-01)
We studied the effects of inhibitors of ornithine decarboxylase (ODC), arginine decarboxylase (ADC) and spermidine synthase (Spd synthase) on organogenesis and the titers of polyamines (PA) and alkaloids in tobacco calli. DL-alpha-diffluromethylarginine (DFMA) and D-arginine (D-Arg), both inhibitors of ADC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service