Skip to Content
Merck
All Photos(1)

Key Documents

297046

Sigma-Aldrich

D-Glucose-1-13C

99 atom % 13C

Synonym(s):

Labeled Glucose, Dextrose-1-13C

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13CC5H12O6
CAS Number:
Molecular Weight:
181.15
Beilstein:
2263682
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

99 atom % 13C

Assay

99% (CP)

form

powder

technique(s)

bio NMR: suitable

storage temp.

room temp

SMILES string

OC[C@H]1O[13CH](O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5-,6?/m1/s1/i6+1

InChI key

WQZGKKKJIJFFOK-USBRANDWSA-N

Looking for similar products? Visit Product Comparison Guide

Application

D-Glucose-1-13C can be used:
  • To predict the primary reaction mechanism in pyrolysis of glucose.
  • To study C−C bond cleavage mechanism in the conversion of labeled glucose into alkanediols using Ni−MgO−ZnO catalyst.
  • In tracer enrichment determination in blood plasma using high-resolution mass spectrometry.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Fleur Sernee et al.
Cell host & microbe, 26(3), 385-399 (2019-09-13)
Parasitic protists belonging to the genus Leishmania synthesize the non-canonical carbohydrate reserve, mannogen, which is composed of β-1,2-mannan oligosaccharides. Here, we identify a class of dual-activity mannosyltransferase/phosphorylases (MTPs) that catalyze both the sugar nucleotide-dependent biosynthesis and phosphorolytic turnover of mannogen. Structural
Catalytic conversion of glucose into alkanediols over nickel-based catalysts: a mechanism study.
Tan Z, et al.
Royal Society of Chemistry Advances, 6(67), 62747-62753 (2016)
Evaluation of primary reaction pathways in thin-film pyrolysis of glucose using 13C labeling and real-time monitoring.
Hutchinson CP and Lee YJ
ACS sustainable chemistry & engineering, 5(10), 8796-8803 (2017)
Ingjye Jiang et al.
Nucleic acids research, 39(20), 8992-9008 (2011-07-21)
Iron-inducible transcription of the ap65-1 gene in Trichomonas vaginalis involves at least three Myb-like transcriptional factors (tvMyb1, tvMyb2 and tvMyb3) that differentially bind to two closely spaced promoter sites, MRE-1/MRE-2r and MRE-2f. Here, we defined a fragment of tvMyb2 comprising
Determination of the Isotopic Enrichment of 13C-and 2H-Labeled Tracers of Glucose Using High-Resolution Mass Spectrometry: Application to Dual-and Triple-Tracer Studies.
Trotzmuller M, et al.
Analytical Chemistry, 89(22), 12252-12260 (2017)

Articles

Isotopic Labeling for NMR Spectroscopy of Biological Solids;

Review the 10 steps of glycolysis in the Embden-Meyerhof-Parnas glycolytic pathway. Easily compare reaction stages and buy the enzymes for your life science research.

Review the 10 steps of glycolysis in the Embden-Meyerhof-Parnas glycolytic pathway. Easily compare reaction stages and buy the enzymes for your life science research.

Review the 10 steps of glycolysis in the Embden-Meyerhof-Parnas glycolytic pathway. Easily compare reaction stages and buy the enzymes for your life science research.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service