Skip to Content
Merck
All Photos(1)

Key Documents

295000

Sigma-Aldrich

Argon

≥99.998%

Synonym(s):

Argon gas

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ar
CAS Number:
Molecular Weight:
39.95
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:
NACRES:
NA.22

vapor density

1.38 (21 °C, vs air)

Quality Level

Assay

≥99.998%

impurities

≤3 ppm oxygen
≤3 ppm water

bp

-185.7 °C (lit.)

mp

-189.2 °C (lit.)

SMILES string

[Ar]

InChI

1S/Ar

InChI key

XKRFYHLGVUSROY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Supplied in a carbon steel lecture bottle with a CGA180M/CGA110F needle valve installed.

Compatible with the following:
  • Aldrich® lecture-bottle station systems
  • Aldrich® lecture-bottle gas regulators

Other Notes

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC

hose barb

Product No.
Description
Pricing

purge valve

Product No.
Description
Pricing

Pictograms

Gas cylinder

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Press. Gas Compr. Gas

Storage Class Code

2A - Gases

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yonas Beyene et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(5), 1584-1591 (2013-01-30)
The Acheulean technological tradition, characterized by a large (>10 cm) flake-based component, represents a significant technological advance over the Oldowan. Although stone tool assemblages attributed to the Acheulean have been reported from as early as circa 1.6-1.75 Ma, the characteristics
Kenta Mizuse et al.
The journal of physical chemistry. A, 117(5), 929-938 (2013-01-22)
We present infrared spectra of nominal water cluster radical cations (H(2)O)(n)(+) (n = 3-8), or to be precise, ion-radical complexes H(+)(H(2)O)(n-1)(OH), with and without an Ar tag. These clusters are closely related to the ionizing radiation-induced processes in water and
Linlin Li et al.
Small (Weinheim an der Bergstrasse, Germany), 11(20), 2429-2436 (2015-01-22)
Herein, the controlled synthesis of 3D hierarchical films on carbon cloth (CC) in a high yield through a hydrothermal process and their high photocatalytic properties are reported. As representative examples, the obtained ZnIn2 S4 /CdIn2 S4 composites are composed of
Andrew Riddell et al.
Methods (San Diego, Calif.), 82, 64-73 (2015-03-10)
Sorting performance can be evaluated with regard to Purity, Yield and/or Recovery of the sorted fraction. Purity is a check on the quality of the sample and the sort decisions made by the instrument. Recovery and Yield definitions vary with
Paul R Renne et al.
Science (New York, N.Y.), 339(6120), 684-687 (2013-02-09)
Mass extinctions manifest in Earth's geologic record were turning points in biotic evolution. We present (40)Ar/(39)Ar data that establish synchrony between the Cretaceous-Paleogene boundary and associated mass extinctions with the Chicxulub bolide impact to within 32,000 years. Perturbation of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service