Skip to Content
Merck
All Photos(1)

Documents

294535

Sigma-Aldrich

meso-Hydrobenzoin

99%

Synonym(s):

meso-1,2-Diphenyl-1,2-ethanediol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(OH)CH(OH)C6H5
CAS Number:
Molecular Weight:
214.26
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

solid

mp

137-139 °C (lit.)

solubility

ethanol: soluble 25 mg/mL, clear, colorless

SMILES string

O[C@H]([C@H](O)c1ccccc1)c2ccccc2

InChI

1S/C14H14O2/c15-13(11-7-3-1-4-8-11)14(16)12-9-5-2-6-10-12/h1-10,13-16H/t13-,14+

InChI key

IHPDTPWNFBQHEB-OKILXGFUSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Desymmetrization of meso-hydrobenzoin using chiral phosphine catalyst has been reported. Conversion of meso-hydrobenzoin to trans-stillbene oxide by treatment with an aryl sulfonyl chloride and aqueous sodium hydroxide has been reported.

Application

meso-Hydrobenzoin has been used in the preparation of trans-methyl meso-hydrobenzoin phosphite.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation and crystal structures of trans-methyl meso-hydrobenzoin phosphite and phosphate.
Newton MG and Campbell BS.
Journal of the American Chemical Society, 96(25), 7790-7797 (1974)
Conversion of meso-Hydrobenzoin with Arylsulfonyl Chlorides and Base to trans-Stilbene Oxide and to 1, 1-Diphenyl-2-(p-toluenesulfonyloxy) ethylene1.
Curtin DY and Hendrickson YG.
The Journal of Organic Chemistry, 21(11), 1260-1263 (1956)
E Vedejs et al.
The Journal of organic chemistry, 69(4), 1389-1392 (2004-02-14)
The desymmetrization of meso-hydrobenzoin is described using chiral phosphine catalysts 8b-d and 9-11. The best enantioselectivity at room temperature was obtained with the newly synthesized phospholane 8c and benzoic anhydride, but the reaction is very slow. Much faster reactions, but

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service