Skip to Content
Merck
meso-Hydrobenzoin 99%

meso-Hydrobenzoin

meso-Hydrobenzoin 99%
Synonym(s):
meso-1,2-Diphenyl-1,2-ethanediol
Linear Formula:
C6H5CH(OH)CH(OH)C6H5
CAS Number:
Molecular Weight:
214.26

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing