Skip to Content
Merck
All Photos(1)

Documents

29226

Sigma-Aldrich

Undecafluorohexanoic acid

≥97.0% (T)

Synonym(s):

Perfluorocaproic acid, Perfluorohexanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3(CF2)4COOH
CAS Number:
Molecular Weight:
314.05
Beilstein:
1805852
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97.0% (T)

solubility

water: insoluble

density

1.759 g/mL at 20 °C (lit.)

SMILES string

OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C6HF11O2/c7-2(8,1(18)19)3(9,10)4(11,12)5(13,14)6(15,16)17/h(H,18,19)

InChI key

PXUULQAPEKKVAH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Undecafluorohexanoic acid, a volatile ion-pair reagent, has been used as mobile phase for LC-MS analysis of highly polar sulfonium pseudo-sugar constituents neosalacinol and neokotalanol, potential α-glucosidase inhibitors isolated from ayurvedic traditional medicine Salacia species.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Osamu Muraoka et al.
Journal of natural medicines, 65(1), 142-148 (2010-10-29)
A quantitative analytical method for the highly polar sulfonium pseudo-sugar constituents neosalacinol (3) and neokotalanol (4), another two potent α-glucosidase inhibitors isolated from Ayurvedic traditional medicine Salacia species, was developed by employing an ion pair reagent upon chromatographic separation. The
Qian S Liu et al.
Environmental science & technology, 51(17), 10173-10183 (2017-07-27)
Per- and polyfluoroalkyl substances (PFASs) are a global concern because of their ubiquitous occurrence and high persistence in human blood, and increasing amounts of unidentified fluorinated compounds are now becoming new exposure issues. This study aims to investigate the structure-related
Abdullah Akhdhar et al.
Talanta, 209, 120466-120466 (2020-01-02)
The determination of total fluorine content using high-resolution graphite furnace continuum source molecular absorption spectrometry (HR- MAS) has been employed in a variety of samples for over 10 years. However, most of the samples analysed by HR- MAS are rich
Lei Wang et al.
Journal of chromatography. A, 1515, 45-53 (2017-08-15)
Ionic liquids have been used to efficiently extract a wide range of polar and nonpolar organic contaminants from water. In this study, imidazole ionic liquids immobilized on silica gel were synthesized through a chemical bonding method, and the immobilized dodecylimidazolium
Yi-Bo Hu et al.
Water research, 140, 148-157 (2018-04-29)
An autocatalytic system, permanganate-ultrasonic (PM-US) system, was applied to degrade perfluorooctanoic acid (PFOA) in aqueous solutions. After a 120-min ultrasonication, a PM dosage of 6 mM increased the pseudo first-order rate constant (k1) for PFOA decomposition from 3.5 × 10-3 to 13.0 × 10-3 min-1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service