Skip to Content
Merck
All Photos(1)

Key Documents

294209

Sigma-Aldrich

Potassium nonafluoro-1-butanesulfonate

98%

Synonym(s):

Nonafluoro-1-butanesulfonic acid potassium salt, Potassium nonaflate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3(CF2)3SO3K
CAS Number:
Molecular Weight:
338.19
Beilstein:
5200982
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

>300 °C (lit.)

solubility

water: soluble 100 mg/mL, clear, colorless

SMILES string

[K+].[O-]S(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C4HF9O3S.K/c5-1(6,3(9,10)11)2(7,8)4(12,13)17(14,15)16;/h(H,14,15,16);/q;+1/p-1

InChI key

LVTHXRLARFLXNR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An Improved Total Synthesis of Tubercidin.
Huang H, et al.
Chinese Journal of Organic Chemistry, 34, 1358-1363 (2014)
Xiaojuan You et al.
Molecules (Basel, Switzerland), 23(7) (2018-07-14)
The present study investigated the adsorptive and enzymatic removal of aniline blue dye (AB) from aqueous solution using waxy riceprocessing waste (RW), peanut shell (PS), microbial waste of Aspergillus niger (MW) as low cost adsorbents, and laccase (Lac) as a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service